GE17-3717-47
HiScreen™ Capto™ S ImpAct
Cytiva 17-3717-47
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
23151817
NACRES:
NA.56
Recommended Products
ligand
sulfonate group
manufacturer/tradename
Cytiva 17-3717-47
matrix
highly cross-linked agarose with dextran surface extender
avg. part. size
90 μm (d50V)
cleaning
3-14
working range
4-12
Related Categories
General description
HiScreen Capto S ImpAct (4.7 ml) are prepacked columns for small scale purifications, as well as optimization of methods and parameters, such as sample load and binding conditions. HiScreen Capto S ImpAct columns provide fast, reproducible and easy separations in a convenient format. The columns are best utilized when they are connected to liquid chromatography systems such as ÄKTA systems.
Capto S ImpAct is a BioProcess chromatography medium developed for large-scale purification of biopharmaceutical proteins. Based on the well-established Capto base matrix, it provides many opportunities for improved productivity and straightforward process development. This chromatography medium (resin) is available in a range of different bulk pack sizes and convenient prepacked formats for easy scale-up and process development. As a member of the BioProcess media range, Capto S ImpAct meets industrial demands with security of supply and comprehensive technical and regulatory support.
Capto S ImpAct is a BioProcess chromatography medium developed for large-scale purification of biopharmaceutical proteins. Based on the well-established Capto base matrix, it provides many opportunities for improved productivity and straightforward process development. This chromatography medium (resin) is available in a range of different bulk pack sizes and convenient prepacked formats for easy scale-up and process development. As a member of the BioProcess media range, Capto S ImpAct meets industrial demands with security of supply and comprehensive technical and regulatory support.
Features and Benefits
HiScreen Capto S ImpAct columns are prepacked with Capto S ImpAct cation exchange chromatography medium, and are part of our process development platform. The columns are an excellent choice for method optimization and parameter screening.
10 cm bed height of HiScreen columns is designed to allow method optimization and parameter screening.
10 cm bed height of HiScreen columns is designed to allow method optimization and parameter screening.
- Small bed volume gives fast results and minimal sample/buffer consumption.
- Easily connected in series to achieve 20 cm bed height.
- Reproducible results, scalable to BioProcess columns packed with the same chromatography media using the same linear fluid velocity.
- Prepacked with Capto S ImpAct, a strong cation exchange BioProcess medium, designed for immediate purification and polishing of a wide range of biomolecules, especially monoclonal antibodies.
Storage and Stability
Store at 4 to 30 °C (20% Ethanol)
Analysis Note
To view the Certificate of Analysis for this product, please visit www.cytiva.com.
Legal Information
Capto is a trademark of Cytiva
HiScreen is a trademark of Cytiva
related product
Product No.
Description
Pricing
Storage Class Code
3 - Flammable liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service