GE17-3717-51
HiTrap® Capto™ S ImpAct 5x1mL
Cytiva 17-3717-51, pack of 5x1ml
Synonym(s):
HITRAP® CAPTO™ S IMPACT
About This Item
Recommended Products
ligand
(-SO3)
form
prepacked column
crosslinking
cross-linked
manufacturer/tradename
Cytiva 17-3717-51
storage condition
(0.2 M sodium acetate in 20% ethanol)
parameter
(Flow velocity: 220 - 600 cm/h)
-44 psi max. pressure
1-4 mL/min flow rate
technique(s)
liquid chromatography (LC): suitable
matrix
high-flow agarose
matrix active group
Polymeric Pyrrolidone and Sulfonate
avg. part. size
50 μm (d50V)
cleaning in place
4-12
working range
3-14
capacity
binding capacity(>85 mg BSA/mL medium, >90 mg lysozyme/ml medium, >100 mg IgG/ ml medium)
separation technique
cation exchange
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Capto S ImpAct is a BioProcess chromatography medium developed for large-scale purification of biopharmaceutical proteins. Based on the well-established Capto base matrix, it provides many opportunities for improved productivity and straightforward process development. This chromatography medium (resin) is available in a range of different bulk pack sizes and convenient prepacked formats for easy scale-up and process development. As a member of the BioProcess media range, Capto S ImpAct meets industrial demands with security of supply and comprehensive technical and regulatory support.
Features and Benefits
- Convenient prepacked column for fast, reproducible, and easy small-scale purifications.
- High binding capacity, > 100 mg MAb/mL medium.
- Efficient aggregate removal at high load of monoclonal antibodies.
- High-resolution intermediate purification and polishing based on the well-established Capto platform.
- Flexibility of design thanks to a large operational window of flow rates and bed heights for easy optimization and scaling.
Storage and Stability
Analysis Note
Legal Information
related product
Storage Class Code
3 - Flammable liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Protocols
This page covers detailed information on cleaning procedures and recommended flow for column cleaning.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service