Skip to Content
Merck
All Photos(2)

Key Documents

G9518

Sigma-Aldrich

Gemfibrozil

Synonym(s):

2,2-Dimethyl-5-(2,5-dimethylphenoxy)pentanoic acid, 2,2-Dimethyl-5-(2,5-xylyloxy)valeric acid, 5-(2,5-Dimethylphenoxy)-2,2-dimethylpentanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H22O3
CAS Number:
Molecular Weight:
250.33
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

powder

Quality Level

SMILES string

Cc1ccc(C)c(OCCCC(C)(C)C(O)=O)c1

InChI

1S/C15H22O3/c1-11-6-7-12(2)13(10-11)18-9-5-8-15(3,4)14(16)17/h6-7,10H,5,8-9H2,1-4H3,(H,16,17)

InChI key

HEMJJKBWTPKOJG-UHFFFAOYSA-N

Gene Information

human ... PPARA(5465)

Looking for similar products? Visit Product Comparison Guide

General description

Gemfibrozil is a fibric acid derivative and a hypolipidemic drug. Gemfibrozil serves as a ligand for PPARα (peroxisome proliferator-activated receptor α).

Application

Gemfibrozil has been used to to study its effects on cell cycle progression in yeast. It has been used to study the effects of fibrates on cell proliferation and gene expression in human cell lines.

Biochem/physiol Actions

Gemfibrozil selectively increases Apolipoprotein A-I levels. In yeast cells, application of gemfibrozil delays the start of DNA replication. It is used as a therapeutic agent for dyslipidemia.

Features and Benefits

This compound is a featured product for ADME Tox research. Click here to discover more featured ADME Tox products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katerina Cizkova et al.
Lipids in health and disease, 15(1), 164-164 (2016-09-24)
Fibrates are widely used hypolipidemic drugs, which serve as ligand of peroxisome proliferator-activated receptor α (PPARα). Recently, they have also been considered as potential anticancer agents. We studied effect of fibrates treatment on cell proliferation, expression of CYP2J2 and concomitant
Izabela Zakrocka et al.
Life (Basel, Switzerland), 13(11) (2023-11-25)
Kidney dysfunction significantly increases the cardiovascular risk, even in cases of minor functional declines. Hypertriglyceridemia is the most common lipid abnormality reported in patients with kidney disorders. PPAR-α (peroxisome proliferator-activated receptor-α) agonists called fibrates are the main agents used to
Benjamin Allcock et al.
Cells, 12(19) (2023-10-13)
It is widely recognised that cells respond to their microenvironment, which has implications for cell culture practices. Growth cues provided by 2D cell culture substrates are far removed from native 3D tissue structure in vivo. Geometry is one of many
Manthena V S Varma et al.
Pharmaceutical research, 29(10), 2860-2873 (2012-05-29)
To develop physiologically based pharmacokinetic (PBPK) model to predict the pharmacokinetics and drug-drug interactions (DDI) of pravastatin, using the in vitro transport parameters. In vitro hepatic sinusoidal active uptake, passive diffusion and canalicular efflux intrinsic clearance values were determined using
A M Filppula et al.
Clinical pharmacology and therapeutics, 94(3), 383-393 (2013-05-10)
Cytochrome P450 (CYP) 3A4 is considered the most important enzyme in imatinib biotransformation. In a randomized, crossover study, 10 healthy subjects were administered gemfibrozil 600 mg or placebo twice daily for 6 days, and imatinib 200 mg on day 3, to study

Articles

Lipoproteins package cholesterol for transport in plasma, essential for lipid transport and cellular function in the body.

Lipoproteins package cholesterol for transport in plasma, essential for lipid transport and cellular function in the body.

Lipoproteins package cholesterol for transport in plasma, essential for lipid transport and cellular function in the body.

Lipoproteins package cholesterol for transport in plasma, essential for lipid transport and cellular function in the body.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service