Skip to Content
Merck
All Photos(2)

Key Documents

G6532

Sigma-Aldrich

Anti-GroEL antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-GroEL, GroEL Antibody - Anti-GroEL antibody produced in rabbit - Tested Applications: immunoblotting 1:80,000,indirect ELISA 1:40,000-1:80,000 - Sigma Aldrich G6532, Groel Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

E. coli

technique(s)

indirect ELISA: 1:40,000-1:80,000
western blot: 1:80,000

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Escherichia coli ... groEL(913705) , groEL(913705)
Escherichia coli K12 ... Grol(948665)

Looking for similar products? Visit Product Comparison Guide

General description

GroEL protein belongs to the folding chaperonin family. It is a 58.3 kDa protein with apical, intermediate and equatorial functional domains.

Specificity

The antibody does not cross-react with GroES and bovine HSP 70.

Immunogen

purified recombinant GroEL produced in E. coli.

Application

Anti-GroEL antibody produced in rabbit has been used as positive control in histology and immunohistochemistry of Dirofilaria immitis, nematode worms and in the western blot analysis of the HeLa cells.

Biochem/physiol Actions

GroEL, together with GroES, in the presence of ATP, assists in protein folding. Co-expression of aldehyde dehydrogenase with GroEL/GroES promotes folding and higher production of protein in soluble form. GroES and GroEL bind to the protein substrate, enclosing it in a cage like fold. The target protein after folding gets dissociated from the cage and is accompanied with hydrolysis of adenosine triphosphate (ATP) to adenosine diphosphate (ADP).

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rosa Morra et al.
mBio, 9(1) (2018-02-01)
The apparent mislocalization or excretion of cytoplasmic proteins is a commonly observed phenomenon in both bacteria and eukaryotes. However, reports on the mechanistic basis and the cellular function of this so-called "nonclassical protein secretion" are limited. Here we report that
Chaperonin GroESL mediates the protein folding of human liver mitochondrial aldehyde dehydrogenase in Escherichia coli.
Lee KH, et al.
Biochemical and Biophysical Research Communications, 298(2), 216-224 (2002)
Verena Kohler et al.
Nucleic acids research, 46(17), 9201-9219 (2018-07-31)
The dissemination of multi-resistant bacteria represents an enormous burden on modern healthcare. Plasmid-borne conjugative transfer is the most prevalent mechanism, requiring a type IV secretion system that enables bacteria to spread beneficial traits, such as resistance to last-line antibiotics, among
Immunohistochemical/immunogold detection and distribution of the endosymbiont Wolbachia of Dirofilaria immitis and Brugia pahangi using a polyclonal antiserum raised against WSP (Wolbachia surface protein).
Kramer L, et al.
Parasitology Research, 89(5), 381-386 (2003)
The chlamydial periplasmic stress response serine protease cHtrA is secreted into host cell cytosol.
Wu X, et al.
BMC Microbiology, 11(1), 87-87 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service