Skip to Content
Merck
All Photos(1)

Documents

G5168

Sigma-Aldrich

(Z)-Guggulsterone

≥89% (HPLC), powder

Synonym(s):

(17Z)-Pregna-4,17(20)-diene-3,16-dione, 4,17(20)-trans-Pregnadiene-3,16-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H28O2
CAS Number:
Molecular Weight:
312.45
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥89% (HPLC)

form

powder

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤5.4% E-form

color

light yellow

solubility

DMSO: 5 mg/mL

application(s)

forensics and toxicology
veterinary

storage temp.

2-8°C

SMILES string

[H][C@@]12CCC3=CC(=O)CC[C@]3(C)[C@@]1([H])CC[C@]4(C)\C(=C\C)C(=O)C[C@@]24[H]

InChI

1S/C21H28O2/c1-4-16-19(23)12-18-15-6-5-13-11-14(22)7-9-20(13,2)17(15)8-10-21(16,18)3/h4,11,15,17-18H,5-10,12H2,1-3H3/b16-4+/t15-,17+,18+,20+,21-/m1/s1

InChI key

WDXRGPWQVHZTQJ-OSJVMJFVSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

(Z)-Guggulsterone is a natural product that lowers cholesterol due to its function as an antagonist ligand for the bile acid receptor. (Z)-Guggulsterone is a nuclear hormone receptor that regulates the transcription of several genes involved in cholesterol metabolism and plays a role in cholesterol level regulation. (Z)-Guggulsterone is also a selective farnesoid X receptor (FXR) modulator.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Dong-Won Ahn et al.
Pancreas, 41(7), 1048-1057 (2012-04-20)
Guggulsterone is a dietary plant sterone possessing therapeutic potential against cancers. However, the antitumor effect of this natural compound on pancreatic cancer has not been determined yet. This study was designed to investigate the therapeutic efficacy of guggulsterone in pancreatic
Rodolfo Gonzalez et al.
Scientific reports, 3, 1463-1463 (2013-03-16)
New small molecules that regulate the step-wise differentiation of human pluripotent stem cells into dopaminergic neurons have been identified. The steroid, guggulsterone, was found to be the most effective inducer of neural stem cells into dopaminergic neurons. These neurons are
R S Bhatta et al.
Biomedical chromatography : BMC, 25(9), 1054-1060 (2011-01-27)
A sensitive and selective liquid chromatography/tandem mass spectrometric method was developed for simultaneous determination of E- and Z-guggulsterone isomers (antihyperlipidemic drug) in rabbit plasma. Both the isomers were resolved on a Symmetry-Shield C(18) (5 µm, 4.6 × 150 mm) column, using gradient
Y T Kamal et al.
Biomedical chromatography : BMC, 26(10), 1183-1190 (2012-01-04)
An attempt has been made to develop and validate a simultaneous HPLC method for novel approach of drug release via oil-in-water (o/w) nanoemulsion formulation and Habb-e-Khardal Unani tablet containing piperine and guggul sterones E and Z as main ingredients. Nanoemulsion
Jeung-Hyun Koo et al.
International journal of molecular medicine, 30(4), 974-978 (2012-07-17)
In the present study, we investigated the effect of guggulsterone on melanogenesis in B16 melanoma cells and elucidated its possible mechanism of action. The effects of guggulsterone on melanogenesis were determined by assaying melanin synthesis and cellular tyrosinase activity in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service