Skip to Content
Merck
All Photos(3)

Key Documents

N5757

Sigma-Aldrich

α-Naphthoflavone

≥98%

Synonym(s):

alpha-Naphthoflavone, 7,8-Benzoflavone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H12O2
CAS Number:
Molecular Weight:
272.30
Beilstein:
210494
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

≥98%

form

powder

technique(s)

titration: suitable

color

white to yellow

mp

153-157 °C (lit.)

storage temp.

2-8°C

SMILES string

O=C1C=C(Oc2c1ccc3ccccc23)c4ccccc4

InChI

1S/C19H12O2/c20-17-12-18(14-7-2-1-3-8-14)21-19-15-9-5-4-6-13(15)10-11-16(17)19/h1-12H

InChI key

VFMMPHCGEFXGIP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Min Ji Kim et al.
Environmental health perspectives, 120(4), 508-514 (2012-01-21)
Epidemiological studies emphasize the possible role of persistent organic pollutants (POPs) in obesity and the metabolic syndrome. These pollutants are stored in adipose tissue (AT). Our aim was to study the effects of POPs on human adipose cells and rodent
Po-Lin Liao et al.
Experimental dermatology, 21(7), 546-548 (2012-06-22)
Ageing is a complex and multifactorial process resulting in several functional and aesthetic changes to the skin. We found that α-Naphthoflavone (α-NF) concentration-dependently induced pro-collagen type I protein expression and inhibited MMP-1 protein expression, in both normal and UVB-irradiated cells.
Iain A Murray et al.
Molecular pharmacology, 79(3), 508-519 (2010-12-04)
We have characterized previously a class of aryl hydrocarbon receptor (AHR) ligand termed selective AHR modulators (SAhRMs). SAhRMs exhibit anti-inflammatory properties, including suppression of cytokine-mediated acute phase genes (e.g., Saa1), through dissociation of non-dioxin-response element (DRE) AHR activity from DRE-dependent
Barbara M Zietek et al.
SLAS discovery : advancing life sciences R & D, 23(3), 283-293 (2017-12-22)
With early assessment of inhibitory properties of drug candidates and their circulating metabolites toward cytochrome P450 enzymes, drug attrition, especially later in the drug development process, can be decreased. Here we describe the development and validation of an at-line nanofractionation
Ruijuan Liu et al.
Frontiers in pharmacology, 9, 1264-1264 (2018-11-22)
Corynoline (CRL), an isoquinoline alkaloid, is the major constituent derived from Corydalis bungeana Herba, which is a well-known Chinese herbal medicine widely used in many prescriptions. The purpose of this study was to comprehensively investigate the metabolism and bioactivation of

Articles

Antioxidants protect biological systems from oxidative damage produced by oxygen-containing free radicals and from redoxactive transition metal ions such as iron, copper, and cadmium.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service