Skip to Content
Merck
All Photos(2)

Key Documents

G2526

Sigma-Aldrich

Gel Loading Buffer

for NA electrophoresis

Synonym(s):

6X DNA loading buffer, 6X DNA loading dye, 6X DNA loading solution, agarose gel loading buffer, agarose gel loading dye, agarose gel loading solution, sample loading buffer, sample loading dye, sample loading solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161703
NACRES:
NA.31

grade

for molecular biology

Quality Level

form

liquid

foreign activity

RNAse, none detected

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

General description

Gel loading buffer is used as a tracking dye during electrophoresis. The dye has a slight negative charge and will migrate the same direction as DNA, allowing the user to monitor the progress of molecules moving through the gel. The rate of migration varies with gel composition. Dilute 1:3 to 1:6 with sample before loading.

Application

Gel Loading Buffer has been used for loading polymerase chain reaction (PCR) amplified intergenic spacer region (ISR) sequence samples, PCR amplified DNA samples of the intestinal mucosa, Trypanosoma sp DNA on agarose gel for electrophoresis.It is suitable for use with agarose or non-denaturing polyacrylamide gel electrophoresis (PAGE), which may be part of Northern and Southern blot hybridization procedures.

Biochem/physiol Actions

Gel loading buffer is especially formulated for non-denaturing polyacrylamide and agarose gel electrophoresis of nucleic acids. The gel loading solution contains a bromophenol blue tracking dye and sucrose to add density and facilitate sample loading. EDTA has been included to inhibit nucleases that require divalent cations and SDS has been added to help dissociate DNA-protein complexes which can interfere with electrophoresis.

Components

Gel loading buffer contains 0.05% bromophenol blue , 40% sucrose, 0.1M EDTA (pH 8.0) and 0.5% SDS.

Other Notes

If crystals form, dissolve them by heating the solution to 65C for 10 minutes.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Validation of a polymerase chain reaction assay for monitoring the therapeutic efficacy of diminazene aceturate in trypanosome-infected sheep
Bengaly Z, et al.
Veterinary Parasitology, 96(2), 101-113 (2001)
Identification of dairy lactic acid bacteria by tRNAAla-23S rDNA-RFLP
Mancini A, et al.
Journal of Microbiological Methods, 91(3), 380-390 (2012)
Comparison of diagnostic techniques for porcine proliferative enteropathy (Lawsonia intracellularis infection)
Huerta B, et al.
Journal of Comparative Pathology, 129(2-3), 179-185 (2003)

Related Content

Nancy-520 for DNA Detection and Quantitation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service