Skip to Content
Merck
All Photos(1)

Key Documents

G1767

Sigma-Aldrich

Glycogen from mussel, Mytilus genus

for DNA precipitations

Synonym(s):

DNA and RNA precipitation

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.52

grade

for molecular biology

Quality Level

form

liquid

packaging

vial of 1 mL

concentration

19-22 mg/mL

storage temp.

−20°C

InChI

1S/C24H42O21/c25-1-5-9(28)11(30)16(35)22(41-5)39-4-8-20(45-23-17(36)12(31)10(29)6(2-26)42-23)14(33)18(37)24(43-8)44-19-7(3-27)40-21(38)15(34)13(19)32/h5-38H,1-4H2/t5-,6-,7-,8-,9-,10-,11+,12+,13-,14-,15-,16-,17-,18-,19-,20-,21+,22+,23-,24-/m1/s1

InChI key

BYSGBSNPRWKUQH-UJDJLXLFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Glycogen is a branched polymer of glucose synthesized by animal cells for energy storage and release. It is constructed of predominantly α1→4 glycosidic bonds with branches created through α1→6 glycosidic bonds.
Glycogen is a branched polymer of glucose that coprecipitates nucleic acids during isolation and purification.

Application

Glycogen from Mytilus genus (Blue mussel) has been used


  • as a coprecipitant in the isolation of RNA from hepatocytes and yeast cells.
  • as a reference standard in calibration curve generation to quantify hepatic glycogen content.
  • as a carrier molecule for DNA and RNA, replacing tRNA or sonicated DNA
  • for the precipitation of DNA in the presence of ammonium acetate and ethanol or isopropanol
  • for recovery of tRNA, yeast RNA, sonicated DNA and oligonucleotides as short as 8 base pairs

Biochem/physiol Actions

Glycogen levels in Mytilus genus shows variation during seasonal changes. It is a crucial stored energy reserve for Mussels and is utilized during gametogenesis. Glycogen levels dictate the reproductive capacity and development of mussels larva. Glycogen aids in the coprecipitation of nucleic acids during isolation and purification.

Features and Benefits

  • product provided is 1 mL total volume
  • precipitates even small amounts of DNA and RNA present in dilute solutions
  • does not interfere with OD 260/280 readings

Principle

Glycogen is an inert carrier that traps nucleic acids. It forms a precipitate with DNA/RNA while being insoluble in ethanol solution. Due to the manufacturing process, there is low risk of contaminating nucleic acids in the glycogen. This product is tested for DNase, RNase, and NICKase, but not nucleic acids.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Regulation of gluconeogenesis by Kruppel-like factor 15
Gray S, et al.
Cell Metabolism, 5(4), 305-312 (2007)
Energy storage and reproduction in mussels, Mytilus galloprovincialis: the influence of diet quality
Fearman JA, et al.
Journal of shellfish research., 28(2), 305-312 (2009)
Tri-methylation of histone H3 lysine 4 facilitates gene expression in ageing cells
Cruz C, et al.
eLife, 7, e34081-e34081 (2018)
Biomass composition of blue mussels, Mytilus edulis, is affected by living site and species of ingested microalgae
Pleissner D, et al.
ISRN zoology, 2012 (2012)
Ryan J Farr et al.
PLoS pathogens, 17(7), e1009759-e1009759 (2021-07-29)
The host response to SARS-CoV-2 infection provide insights into both viral pathogenesis and patient management. The host-encoded microRNA (miRNA) response to SARS-CoV-2 infection, however, remains poorly defined. Here we profiled circulating miRNAs from ten COVID-19 patients sampled longitudinally and ten

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service