Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Glycogen from mussel, Mytilus genus
CAS Number:
9005-79-2
EC Number:
232-683-8
Products
Product Category
carbohydrates (1)
polysaccharides (1)
Brand
Sigma-Aldrich (2)
Color
Physical Form
Purity
Quality Segment
Package Size
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
G1767
Glycogen from mussel, Mytilus genus,
for DNA precipitations
Expand
View Pricing
G1508
Glycogen from mussel, Mytilus genus,
≥85% anhydrous basis (enzymatic)
Expand
View Pricing