Skip to Content
Merck
All Photos(3)

Documents

G1546

Sigma-Aldrich

Anti-Green Fluorescent Protein (GFP) antibody, Mouse monoclonal

clone GSN149, purified from hybridoma cell culture

Synonym(s):

Mouse Anti-GFP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

GSN149, monoclonal

form

buffered aqueous solution

packaging

antibody small pack of 25 μL

technique(s)

indirect ELISA: suitable
western blot: 1-2 μg/mL using extracts of cells expressing GFP fusion proteins

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Monoclonal Anti-Green Fluorescent Protein (GFP) (mouse IgG1 isotype) is derived from the hybridoma GSN149 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a synthetic peptide of the Green fluorescent protein from jellyfish Aequorea victoria. GFP is a 27 kDa protein (238 amino acids) that can produce a strong green fluorescence without the need for a substrate. It absorbs light maximally at 395 nm and emits a bright green fluorescence with a peak at 509 nm. The GFP chromophore is formed through cyclization and oxidation of an internal tripeptide motif (Ser65, Gly69 and Tyr66).
The antibody reacts specifically with GFP-fusion proteins.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Monoclonal Anti-Green Fluorescent Protein (GFP) antibody produced in mouse was used in surface plasmon resonance (SPR) gold chip analysis to study the cysteine- and domain-mediated immobilization of enhanced GFP model protein.

Biochem/physiol Actions

GFP is implicated in protein detection and localization in living cells, as well as gene expression monitoring in prokaryotes and eukaryotes. It detects protein-protein interactions in living cells.
Green fluorescent protein, produced in the jellyfish, Aequorea victoria, acts as secondary fluorescent protein in an energy transfer reaction to produce green light. It ability to produce a highly visible, internal fluorophore is widely applicable as marker for gene expression and to produce chimeric proteins to understand protein biochemistry.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kyoungsook Park et al.
The Analyst, 136(12), 2506-2511 (2011-04-27)
Here we report an effective method for protein immobilization on a surface plasmon resonance (SPR) gold chip, describing the combination of cysteine- and oligomerization domain-mediated immobilization of enhanced green fluorescent protein (EGFP) as a model protein for the purpose of
The dynamic microbe: green fluorescent protein brings bacteria to light
Southward CM and Surette MG
Molecular Microbiology, 45(5), 1191-1196 (2002)
GFP: Lighting up life.
Martin Chalfie
Proceedings of the National Academy of Sciences of the United States of America, 106(25), 10073-10080 (2009-06-26)
Visual screening of microspore-derived transgenic barley (Hordeum vulgare L.) with green-fluorescent protein
Carlson AR, et al.
Plant Cell Reports, 20(4), 331-337 (2001)
Petra C Boevink et al.
Nature communications, 7, 10311-10311 (2016-01-30)
Plant pathogens deliver effectors to alter host processes. Knowledge of how effectors target and manipulate host proteins is critical to understand crop disease. Here, we show that in planta expression of the RXLR effector Pi04314 enhances leaf colonization by Phytophthora

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service