Skip to Content
Merck
All Photos(5)

Documents

MAB3580

Sigma-Aldrich

Anti-Green Fluorescent Protein Antibody

Chemicon®, from mouse

Synonym(s):

GFP, eGFP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

monoclonal

species reactivity (predicted by homology)

all

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

target post-translational modification

unmodified

General description

The Green Fluorescent Protein (GFP) from the jellyfish Aequorea victoria is used as a fluorescent indicator for monitoring gene expression in a variety of cellular systems, including living organisms and fixed tissues. Unlike other bioluminescent reporters, GFP fluoresces in the absence of substrates, cofactors, or other intrinsic or extrinsic proteins. Purified GFP is a 27 kDa monomer consisting of 238 amino acids and emits green light (emission maximum at 509 nm) when excited with blue or UV light.

Specificity

Reacts with Green Fluorescent Protein (GFP). The antibody reacts with various forms of GFP including eGFP including those found in vectors supplied by Clonetech and Invitrogen. There is not cross-reactivity to RFP. YFP has not been tested. This antibody works well for immunohistochemistry.

Immunogen

GFP-BSA

Application

Detect Green Fluorescent Protein using this Anti-Green Fluorescent Protein Antibody validated for use in ELISA, IC, IH & WB.
Western blot: 1:2,500-1:5,000. Recommended dilution buffer is PBS containing 1 mg/mL BSA.

Immunocytochemistry: 1:500-1:1,000

Immunohistochemistry: 1:500-1:1,000 on paraformaldehyde fixed tissues with an overnight incubation using Nickel enhanced DAB for detection. Suggested permeabilization method is 0.5% Triton X-100. Recommended blocking buffer is PBS containing BSA and serum from the host of the secondary antibody. Recommended dilution buffer is PBS containing 1 mg/mL of BSA.

ELISA: 1:1,000-1:2,000 in a direct assay.

Optimal working dilutions must be determined by the end user.

Target description

MW for GFP is 27 kDa Dependent upon the molecular weight of the fusion protein being detected.

Physical form

Format: Purified
Liquid in PBS containing 1 mg/mL BSA. Contains no preservative.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Drosophila Dpp morphogen movement is independent of dynamin-mediated endocytosis but regulated by the glypican members of heparan sulfate proteoglycans.
Belenkaya, TY; Han, C; Yan, D; Opoka, RJ; Khodoun, M; Liu, H; Lin, X
Cell null
Multiple phenotypes resulting from a mutagenesis screen for pharynx muscle mutations in Caenorhabditis elegans.
Ferrier, A; Charron, A; Sadozai, Y; Switaj, L; Szutenbach, A; Smith, PA
Testing null
Magdalena Jasińska et al.
Molecular neurobiology, 53(7), 4701-4712 (2015-09-01)
Mir-132 is a neuronal activity-regulated microRNA that controls the morphology of dendritic spines and neuronal transmission. Similar activities have recently been attributed to matrix metalloproteinase-9 (MMP-9), an extrasynaptic protease. In the present study, we provide evidence that miR-132 directly regulates
Benjamin J Spink et al.
Nature structural & molecular biology, 15(6), 591-597 (2008-05-31)
Myosin VI has challenged the lever arm hypothesis of myosin movement because of its ability to take approximately 36-nm steps along actin with a canonical lever arm that seems to be too short to allow such large steps. Here we
Travis B Lewis et al.
PloS one, 5(9) (2010-09-24)
Gene-based therapy is a new paradigm for the treatment of Parkinson disease (PD) and offers considerable promise for precise targeting and flexibility to impact multiple pathobiological processes for which small molecule agents are not available. Some success has been achieved

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service