Skip to Content
Merck
All Photos(3)

Key Documents

F8791

Sigma-Aldrich

5-Fluoro-5′-deoxyuridine

Synonym(s):

Doxifluridine, 5′dFUrd

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11FN2O5
CAS Number:
Molecular Weight:
246.19
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

SMILES string

C[C@H]1O[C@H]([C@H](O)[C@@H]1O)N2C=C(F)C(=O)NC2=O

InChI

1S/C9H11FN2O5/c1-3-5(13)6(14)8(17-3)12-2-4(10)7(15)11-9(12)16/h2-3,5-6,8,13-14H,1H3,(H,11,15,16)/t3-,5-,6-,8-/m1/s1

InChI key

ZWAOHEXOSAUJHY-ZIYNGMLESA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Doxifluridine is an antitumor agent efficient in tumors, cell lines or in fibroblasts transformed by H-ras or trk oncogenes. Possesses anticachectic activity which is independent of its antiproliferative activity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hitoshi Miyakoshi et al.
Journal of medicinal chemistry, 55(7), 2960-2969 (2012-03-13)
Recently, deoxyuridine triphosphatase (dUTPase) has emerged as a potential target for drug development as part of a new strategy of 5-fluorouracil-based combination chemotherapy. We have initiated a program to develop potent drug-like dUTPase inhibitors based on structure-activity relationship (SAR) studies
G J Peters et al.
International journal of cancer, 54(3), 450-455 (1993-05-28)
Malignant activation of oncogenes ras or trk is implicated in a number of solid tumors and leukemias. We determined the chemosensitivity profile of wild-type mouse NIH-3T3 fibroblasts, and that of NIH-3T3 lines transformed by the H-ras (S2-721) and trk (106-632)
Dengke Pan et al.
Chemical communications (Cambridge, England), 47(3), 908-910 (2010-11-17)
Nearly monodispersed magnetic Fe(3)O(4)@DFUR-LDH submicro particles containing the anticancer agent DFUR were prepared via a coprecipitation-calcination-reconstruction strategy of LDH materials over the surface of Fe(3)O(4) particles, and present well-defined core-shell structure, strong magnetization and obvious magnetically controlled drug delivery and
Yasuko Naganuma et al.
Cancer science, 102(8), 1545-1552 (2011-06-03)
Uterine carcinosarcoma is a highly aggressive gynecological neoplasm that responds poorly to conventional chemotherapy and radiotherapy. Metronomic chemotherapy is accepted as a new approach for cancer treatment, and its underlying mechanism seems to involve the suppression of angiogenesis. However, the
Maria Ait-Tihyaty et al.
Breast cancer research and treatment, 133(1), 217-226 (2011-09-15)
Capecitabine (Xeloda) is a prodrug of 5-FU used in the clinical management of advanced breast cancer. It is metabolized first in the liver by carboxylesterases to generate 5'-deoxy-5-flurocytidine ribose (5'-DFCR), which is subsequently converted to 5'-deoxy-5-fluorouridine ribose (5'-DFUR) by cytidine

Articles

DNA damage and repair mechanism is vital for maintaining DNA integrity. Damage to cellular DNA is involved in mutagenesis, the development of cancer among others.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service