Skip to Content
Merck
All Photos(1)

Key Documents

F3523

Sigma-Aldrich

Monoclonal Anti-Digoxin–FITC antibody produced in mouse

clone DI-22, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Monoclonal Anti-Digoxin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

mouse

conjugate

FITC conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

DI-22, monoclonal

form

buffered aqueous solution

storage condition

protect from light

extent of labeling

3-8 mol fluorochrome per mol protein

technique(s)

dot immunobinding: 1:16 using digoxigenin-labeled biomolecule

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Monoclonal Anti-Digoxin (mouse IgG1 isotype) is derived from the hybridoma produced by the fusion of mouse myeloma cells and splenocytes from an immunized mouse.

Specificity

Antibody is specific for digoxin and shows a high affinity for digoxigenin.

Immunogen

Digoxin-KLH.

Application

Monoclonal Anti-Digoxin-FITC antibody produced in mouse has been used in immuno fluorescence in situ hybridization (FISH) and FISH.

Biochem/physiol Actions

Monoclonal Anti-Digoxin- FITC may be used to detect digoxin-labeled compounds such as oligonucleotides, antibodies or peptides. Labeled compounds and corresponding conjugated antibodies can be used for the detection of viruses and bacterial infections in human diagnostics, oncogenes as tumor markers, histocompatibility antigens in transplantation analytics causative research (e.g., in autoimmune diseases), characterization of lymphoid cell subpopulations (e.g., during treatment of lymphomas), determination of genetic defects or genetic defect predispositions (e.g., Alzheimer′s disease), and nucleic acid diagnostics.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% BSA and 15 mM sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nomar Espinosa Waminal et al.
PloS one, 8(9), e74060-e74060 (2013-09-17)
Monitoring of genetically modified (GM) crops has been emphasized to prevent their potential effects on the environment and human health. Monitoring of the inadvertent dispersal of transgenic maize in several fields and transport routes in Korea was carried out by
Aayushi Uberoi et al.
PLoS pathogens, 12(5), e1005664-e1005664 (2016-06-01)
Human papillomaviruses are causally associated with 5% of human cancers. The recent discovery of a papillomavirus (MmuPV1) that infects laboratory mice provides unique opportunities to study the life cycle and pathogenesis of papillomaviruses in the context of a genetically manipulatable
H3 K79 dimethylation marks developmental activation of the beta-globin gene but is reduced upon LCR-mediated high-level transcription
Sawado T, et al.
Blood, 112(2), 406-414 (2008)
Heterochromatin evolution in Arachis investigated through genome-wide analysis of repetitive DNA
Samoluk SS, et al.
Planta, 249(5), 1405-1415 (2019)
Jeanlin Jourdain et al.
Genome research, 33(6), 957-971 (2023-07-07)
In this paper, we developed a highly sensitive approach to detect interchromosomal rearrangements in cattle by searching for abnormal linkage disequilibrium patterns between markers located on different chromosomes in large paternal half-sib families genotyped as part of routine genomic evaluations.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service