Skip to Content
Merck
All Photos(1)

Key Documents

DIUTPS-RO

Roche

Digoxigenin-11-dUTP, alkali-stable

Synonym(s):

Digoxigenin-11-dUTP, alkali-labile

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105500

Assay

≥85% (HPLC)

Quality Level

form

solution

mol wt

Mr 1090.7

packaging

pkg of 125 μL (11558706910 [1 mM])
pkg of 5 × 125 μL (11570013910 [5x1 mM])
pkg of 25 μL (11093088910 [1 mM])

manufacturer/tradename

Roche

λmax

222 and 292 nm
22430 and 8780

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Digoxigenin-11-dUTP, alkali-stable is provided in 1 mM tetralithium salt, solution.

Application

Digoxigenin-11-dUTP, alkali-stable has been used to label probes for
fluorescent in situ hHybridization (FISH) and polymerase chain reaction (PCR)
Note: When this nucleotide is subjected to 0.1 - 0.5 NaOH at +15 to +25°C, the DIG moiety will remain attached. It is therefore ideal for use in DIG-labeled DNA that needs to survive exposure to alkaline treatment. However, if you intend to make a DIG-labeled probe that can be removed by alkali denaturation (e.g ., during stripping and reprobing of blots), do not use this alkali-stable preparation. Instead, use the alkali-labile form of DIG-11-dUTP.

Biochem/physiol Actions

Digoxigenin (DIG)-11-deoxyuridine triphosphate (dUTP) replaces deoxythymidine triphosphate (dTTP) in the random-primed DNA labeling reaction or in nick translation in a ratio of 35% DIG-11-dUTP and 65% dTTP. DIG-11-dUTP can also replace dTTP in labeling during polymerase chain reaction (PCR) (e.g., with Taq DNA Polymerase), 3′-tailing (with terminal transferase) and cDNA synthesis (e.g., with transcriptor or M-MuLV reverse transcriptase). It is ideal as a substrate for DNA polymerase, Taq DNA polymerase, terminal transferase and reverse transcriptase.

Features and Benefits

Digoxigenin is bound to deoxyuridine triphosphate via an alkali-resistant ether linkage.

Resistance to alkali: The preparation is stable in the presence of 0.1 - 0.5M NaOH at +15 to +25°C.

Quality

Typical analysis: At least 85% DIG-11-dUTP (HPLC, area%).
Function tested by RPL.

Preparation Note

Working solution: We recommend to prepare a deoxynucleotide mix containing dATP, dCTP, dGTP, dTTP (10 mM, each); (e.g., for the preparation of 100 μl nucleotide mix add 10 μl of dATP, dCTP, dGTP, dTTP (each) to 60 μl Water, PCR Grade).
Storage conditions (working solution): A decomposition of approx. 5% may occur within 4 weeks at 15 to 25 °C.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antony W Shermoen et al.
Current biology : CB, 20(23), 2067-2077 (2010-11-16)
Fast, early embryonic cell cycles have correspondingly fast S phases. In early Drosophila embryos, forks starting from closely spaced origins replicate the whole genome in 3.4 min, ten times faster than in embryonic cycle 14 and a hundred times faster
TRF2 and lamin A/C interact to facilitate the functional organization of chromosome ends
Wood AM, et al.
Nature Communications, 5, 5467-5467 (2014)
The Arabidopsis thaliana DSB formation (AtDFO) gene is required for meiotic double-strand break formation
Zhang C, et al.
The Plant Journal, 72 (2012)
Frequency of aneuploidy in in vitro--matured MII oocytes and corresponding first polar bodies in two dairy cattle (Bos taurus) breeds as determined by dual-color fluorescent in situ hybridization
Nicodemo D, et al.
Theriogenology, 73 (2010)
Cecile Meier et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 22(3), 476-487 (2009-01-13)
The Janus kinase 2 (JAK2)-signal transducers and activators of transcription (STAT) pathway plays an important role in hematological malignancies. Mutations and translocations of the JAK2 gene, mapped at 9p24, lead to constitutive activation of JAK2 and its downstream targets. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service