Skip to Content
Merck
All Photos(1)

Key Documents

EHU073901

Sigma-Aldrich

MISSION® esiRNA

targeting human AXIN1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

ACTTCTGGTTTGCCTGCACTGGCTTCAGGAAGCTGGAGCCCTGTGACTCGAACGAGGAGAAGAGGCTGAAGCTGGCGAGAGCCATCTACCGAAAGTACATTCTTGATAACAATGGCATCGTGTCCCGGCAGACCAAGCCAGCCACCAAGAGCTTCATAAAGGGCTGCATCATGAAGCAGCTGATCGATCCTGCCATGTTTGACCAGGCCCAGACCGAAATCCAGGCCACTATGGAGGAAAACACCTATCCCTCCTTCCTTAAGTCTGATATTTATTTGGAATATACGAGGACAGGCTCGGAGAGCCCCAAAGTCTGTAGTGACCAGAGCTCTGGGTCAGGGACAGGGAAGGGCATATCTGGATACCTGCCGACCTTAAATGAAGATGAGGAATGGAAGTGTGACCAGG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu Chen et al.
PloS one, 10(7), e0133115-e0133115 (2015-07-24)
During development, scaffold proteins serve as important platforms for orchestrating signaling complexes to transduce extracellular stimuli into intracellular responses that regulate dendritic spine morphology and function. Axin ("axis inhibitor") is a key scaffold protein in canonical Wnt signaling that interacts
Shiwei Zhou et al.
Pharmaceutics, 13(3) (2021-04-04)
Genetic evidence has indicated that β-catenin plays a vital role in glucose and lipid metabolism. Here, we investigated whether pyrvinium, an anthelmintic agent previously reported as a down-regulator of cellular β-catenin levels, conferred any metabolic advantages in treatment of metabolic
Yongjuan Zhang et al.
Biochemical and biophysical research communications, 513(1), 261-268 (2019-04-08)
Caveolin-1 has been reported to play an important role in the pathogenesis of acute respiratory distress syndrome (ARDS). This study was designed to identify Caveolin-1-interacting proteins to reveal the molecular mechanisms of ARDS. Yeast two-hybrid screening was performed using Caveolin-1
Xiumei Zhang et al.
OncoTargets and therapy, 13, 5819-5830 (2020-07-02)
Evidence describing TMED3 in the context of breast cancer is scarce, and the effect of TMED3 on Wnt/β-catenin signaling in breast cancer has not been reported. The objective of this study was to determine the potential physiological functions and molecular
Dongshao Chen et al.
Cancer management and research, 11, 1349-1362 (2019-02-28)
Characterized by elevated AFP levels in serum, AFP-producing gastric cancer (APGC) is a very special type of gastric cancer (GC) that is difficult to treat and has poor prognosis. However, little is known about the role of AFP in GC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service