Merck
  • Pyrvinium Treatment Confers Hepatic Metabolic Benefits via β-Catenin Downregulation and AMPK Activation.
Pyrvinium Treatment Confers Hepatic Metabolic Benefits via β-Catenin Downregulation and AMPK Activation.
Pharmaceutics (2021-04-04)
Shiwei Zhou, Obinna N Obianom, Jiangsheng Huang, Dong Guo, Hong Yang, Qing Li, Yan Shu
ABSTRACT
Genetic evidence has indicated that β-catenin plays a vital role in glucose and lipid metabolism. Here, we investigated whether pyrvinium, an anthelmintic agent previously reported as a down-regulator of cellular β-catenin levels, conferred any metabolic advantages in treatment of metabolic disorders. Glucose production and lipid accumulation were analyzed to assess metabolic response to pyrvinium in hepatocytes. The expression of key proteins and genes were assessed by immunoblotting and RT-PCR. The in vivo efficacy of pyrvinium against metabolic disorders was evaluated in the mice fed with a high fat diet (HFD). We found that pyrvinium inhibited glucose production and reduced lipogenesis by decreasing the expression of key genes in hepatocytes, which were partially elicited by the downregulation of β-catenin through AXIN stabilization. Interestingly, the AMPK pathway also played a role in the action of pyrvinium, dependent on AXIN stabilization but independent of β-catenin downregulation. In HFD-fed mice, pyrvinium treatment led to improvement in glucose tolerance, fatty liver disorder, and serum cholesterol levels along with a reduced body weight gain. Our results show that small molecule stabilization of AXIN using pyrvinium may lead to improved glucose and lipid metabolism, via β-catenin downregulation and AMPK activation.
Sigma-Aldrich
Anti-Axin-1 Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Mechlorethamine hydrochloride, 98%
Sigma-Aldrich
MISSION® esiRNA, targeting human AXIN1