Skip to Content
Merck
All Photos(1)

Key Documents

EHU016411

Sigma-Aldrich

MISSION® esiRNA

targeting human MAPKAPK5 (2)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GCTGGGAGCTGGAATTAGTGGTCCAGTTAGAGTCTGTGTAAAGAAATCTACTCAAGAACGGTTTGCGCTGAAAATTCTTCTTGATCGTCCAAAAGCTAGAAATGAGGTACGTCTGCACATGATGTGTGCCACACACCCAAACATAGTTCAGATTATTGAAGTGTTTGCTAACAGTGTCCAGTTTCCCCATGAGTCCAGCCCTAGGGCCCGACTCTTAATTGTAATGGAGATGATGGAAGGGGGAGAGCTATTTCACAGAATCAGCCAGCACCGGCACTTTACAGAGAAGCAAGCCAGCCAAGTAACAAAGCAGGCAACTTTGGCTCTGCGGCACTGTCACTTGTTAAACATTGCGCACAGAGACCTCAAGCCTGAAAATCTGCTTTTTAAGGATAACTCTTTGGATGCCCCAGTGAAGTTGTGTGACTTTGGATTTGCCAAGATTGACCAAGGTGACTTGATGACACCCCAGTTCACCCCTTATTATGTAGCACCCCAGGTACTGGA

Ensembl | human accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sherin Ali Nawaito et al.
American journal of physiology. Heart and circulatory physiology, 316(6), H1281-H1296 (2019-03-23)
MK5 is a protein serine/threonine kinase activated by p38, ERK3, and ERK4 MAPKs. MK5 mRNA and immunoreactivity are detected in mouse cardiac fibroblasts, and MK5 haplodeficiency attenuates the increase in collagen 1-α1 mRNA evoked by pressure overload. The present study
Yoonhee Kim et al.
Molecular neurodegeneration, 11, 4-4 (2016-01-14)
The receptor for advanced glycation end products (RAGE) has been found to interact with amyloid β (Aβ). Although RAGE does not have any kinase motifs in its cytosolic domain, the interaction between RAGE and Aβ triggers multiple cellular signaling involved
Natalia Ronkina et al.
PloS one, 10(8), e0136138-e0136138 (2015-08-22)
MK5 (MAPK-activated protein kinase 5) or PRAK (p38-regulated and -activated kinase) are alternative names for a serine/threonine protein kinase downstream to ERK3/4 and p38 MAPK. A previous gene targeting approach for MK5/PRAK (termed here MK5/PRAK-Δex8) revealed a seemingly tumor-suppressive role
Katarzyna Bogucka et al.
eLife, 9 (2020-04-22)
ERK3 is a ubiquitously expressed member of the atypical mitogen activated protein kinases (MAPKs) and the physiological significance of its short half-life remains unclear. By employing gastrointestinal 3D organoids, we detect that ERK3 protein levels steadily decrease during epithelial differentiation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service