Merck
ERK3/MAPK6 controls IL-8 production and chemotaxis.
eLife (2020-04-22)
Katarzyna Bogucka, Malvika Pompaiah, Federico Marini, Harald Binder, Gregory Harms, Manuel Kaulich, Matthias Klein, Christian Michel, Markus P Radsak, Sebastian Rosigkeit, Peter Grimminger, Hansjörg Schild, Krishnaraj Rajalingam
ABSTRACT
ERK3 is a ubiquitously expressed member of the atypical mitogen activated protein kinases (MAPKs) and the physiological significance of its short half-life remains unclear. By employing gastrointestinal 3D organoids, we detect that ERK3 protein levels steadily decrease during epithelial differentiation. ERK3 is not required for 3D growth of human gastric epithelium. However, ERK3 is stabilized and activated in tumorigenic cells, but deteriorates over time in primary cells in response to lipopolysaccharide (LPS). ERK3 is necessary for production of several cellular factors including interleukin-8 (IL-8), in both, normal and tumorigenic cells. Particularly, ERK3 is critical for AP-1 signaling through its interaction and regulation of c-Jun protein. The secretome of ERK3-deficient cells is defective in chemotaxis of neutrophils and monocytes both in vitro and in vivo. Further, knockdown of ERK3 reduces metastatic potential of invasive breast cancer cells. We unveil an ERK3-mediated regulation of IL-8 and epithelial secretome for chemotaxis.
Sigma-Aldrich
MISSION® esiRNA, targeting human MAPK7
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Sigma-Aldrich
Histopaque®-1077, sterile-filtered, density: 1.077 g/mL
Sigma-Aldrich
Mechlorethamine hydrochloride, 98%
Sigma-Aldrich
MISSION® esiRNA, targeting human MAPKAPK5 (2)
Sigma-Aldrich
Lipopolysaccharides from Salmonella enterica serotype typhimurium, suitable for cell culture, BioXtra, γ-irradiated
Sigma-Aldrich
Anti-phospho-ERK3 (pSer189) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
MG-132, A cell-permeable, potent, reversible proteasome inhibitor (Ki = 4 nM).
Sigma-Aldrich
Protease Inhibitor Cocktail Set I, A cocktail of five protease inhibitors that will inhibit a broad range of proteases and esterases. Supplied with a data sheet.