Skip to Content
Merck
All Photos(1)

Documents

D7409

Sigma-Aldrich

2′,3′-Dideoxyguanosine 5′-triphosphate sodium salt

≥90% (HPLC)

Synonym(s):

ddGTP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16N5O12P3
CAS Number:
Molecular Weight:
491.18
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

Quality Level

Assay

≥90% (HPLC)

form

powder

solubility

water: 5 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

[Na+].OP([O-])(=O)OP(O)(=O)OP(O)(=O)OC[C@@H]1CC[C@@H](O1)n2cnc3C(=O)NC(=N)Nc23

InChI

1S/C10H16N5O12P3.Na/c11-10-13-8-7(9(16)14-10)12-4-15(8)6-2-1-5(25-6)3-24-29(20,21)27-30(22,23)26-28(17,18)19;/h4-6H,1-3H2,(H,20,21)(H,22,23)(H2,17,18,19)(H3,11,13,14,16);/q;+1/p-1/t5-,6+;/m0./s1

InChI key

XCOBCPBPAINELA-RIHPBJNCSA-M

Gene Information

human ... HRAS(3265)

Application

2′,3′-Dideoxyguanosine 5′-triphosphate (ddGTP) is use as a chain terminator inhibitor and a inhibitor of telomerase(s).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tadamasa Sasaki et al.
The Plant journal : for cell and molecular biology, 47(5), 802-810 (2006-07-22)
RNA editing is found in various transcripts from land plant chloroplasts. In tobacco chloroplasts, C-to-U conversion occurs at 36 specific sites including two sites identified in this work. Our RNA editing assay system using chloroplast extracts facilitated biochemical analyses of
H Jonckheere et al.
Journal of virological methods, 61(1-2), 113-125 (1996-09-01)
Many bacterial expression systems have been developed to study the reverse transcriptase (RT) of human immunodeficiency virus type 1 (HIV-1). This enzyme exists in the virions as a heterodimer of a 66 kDa (p66) subunit and a 51 kDa (p51)
H Pelemans et al.
Molecular pharmacology, 57(5), 954-960 (2000-04-25)
Trp-229 is part of the non-nucleoside reverse transcriptase inhibitor (NNRTI)-binding pocket of HIV type 1 (HIV-1) reverse transcriptase (RT), and is also part of the "primer grip" of HIV-1 RT. Using site-directed mutagenesis, seven RT mutants were constructed bearing the
H Zhang et al.
Antiviral research, 24(1), 43-57 (1994-05-01)
Two mutants of HIV-1 reverse transcriptase (RT), Tyr-188-->His and Glu-138-->Arg have been prepared and their catalytic properties and sensitivities to inhibitors studied. As compared to wild type RT, a reduction in catalytic efficiency and turn over number was observed, especially
H Jonckheere et al.
The Journal of biological chemistry, 269(41), 25255-25258 (1994-10-14)
Determination of the three-dimensional structure of the human immunodeficiency virus type-1 (HIV-1) reverse transcriptase (RT) has indicated a totally different folding for the 51-kDa subunit (p51) than for the 66-kDa subunit (p66). The polymerase catalytic site is located on the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service