Skip to Content
Merck
All Photos(1)

Key Documents

D5817

Sigma-Aldrich

DMXAA

≥98% (HPLC), solid, apoptosis inducer

Synonym(s):

5,6-Dimethylxanthenone-4-acetic Acid, ASA404, Vadimezan

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H14O4
CAS Number:
Molecular Weight:
282.29
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

DMXAA, ≥98% (HPLC), solid

Quality Level

Assay

≥98% (HPLC)

form

solid

color

light brown

solubility

DMSO: soluble >10 mg/mL

storage temp.

2-8°C

SMILES string

Cc1ccc2C(=O)c3cccc(CC(O)=O)c3Oc2c1C

InChI

1S/C17H14O4/c1-9-6-7-13-15(20)12-5-3-4-11(8-14(18)19)17(12)21-16(13)10(9)2/h3-7H,8H2,1-2H3,(H,18,19)

InChI key

XGOYIMQSIKSOBS-UHFFFAOYSA-N

General description

5,6-dimethylxanthenone-4-acetic acid (DMXAA) is a flavone acetic acid derivative. It acts as a vascular disrupting agent (VDA), which damages tumor vasculature and stimulates an anti-tumor immune response. It stimulates hemorrhagic necrosis.

Application

5,6-dimethylxanthenone-4-acetic acid (DMXAA) has been used to induce type-I IFN signaling in a stimulator of interferon genes (STING) dependent manner. It has also been used to study STING-dependent signaling in the absence of infection.

Biochem/physiol Actions

DMXAA is an apoptosis inducer; anti-vascular.

Features and Benefits

This compound is a featured product for Apoptosis research. Click here to discover more featured Apoptosis products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bruce C Baguley
The Lancet. Oncology, 4(3), 141-148 (2003-03-08)
The vascular endothelium of tumour tissue, which differs in several ways from that of normal tissues, is a potential target for selective anticancer therapy. By contrast with antiangiogenic agents, antivascular agents target the endothelial cells of existing tumour blood vessels
Christina M Buchanan et al.
Clinical science (London, England : 1979), 122(10), 449-457 (2011-12-07)
The flavone acetic acid derivative DMXAA [5,6-dimethylXAA (xanthenone-4-acetic acid), Vadimezan, ASA404] is a drug that displayed vascular-disrupting activity and induced haemorrhagic necrosis and tumour regression in pre-clinical animal models. Both immune-mediated and non-immune-mediated effects contributed to the tumour regression. The
J Guerra et al.
Science advances, 6(21), eaax3333-eaax3333 (2020-06-05)
Inflammation is an essential part of immunity against pathogens and tumors but can promote disease if not tightly regulated. Self and non-self-nucleic acids can trigger inflammation, through recognition by the cyclic GMP-AMP (cGAMP) synthetase (cGAS) and subsequent activation of the
Gary R Martin et al.
Scientific reports, 9(1), 14281-14281 (2019-10-05)
Detection of cytoplasmic DNA by the host's innate immune system is essential for microbial and endogenous pathogen recognition. In mammalian cells, an important sensor is the stimulator of interferon genes (STING) protein, which upon activation by bacterially-derived cyclic dinucleotides (cDNs)
Non-canonical NF-κB Antagonizes STING Sensor-Mediated DNA Sensing in Radiotherapy
Hou Y, et al.
Immunity, 49(3), 490-503 (2018)

Articles

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service