Skip to Content
Merck
All Photos(3)

Key Documents

D5670

Sigma-Aldrich

Sodium deoxycholate monohydrate

BioXtra, ≥99.0% (titration)

Synonym(s):

3α,12α-Dihydroxy-5β-cholanic acid sodium salt, 7-Deoxycholic acid sodium salt, Desoxycholic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H39NaO4 · H2O
CAS Number:
Molecular Weight:
432.57
Beilstein:
3643164
EC Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.25

description

anionic

Quality Level

product line

BioXtra

Assay

≥99.0% (titration)

form

powder

optical activity

[α]20/D +44±2° in H2O(lit.)

mol wt

432.57 g/mol

technique(s)

electrophoresis: suitable
immunoprecipitation (IP): suitable

impurities

Insoluble matter, passes filter test
≤0.5% Cholic acid

pH

7.5-9.5 (20 °C, 0.1 M in H2O)

solubility

H2O: 0.1 M, clear, colorless

anion traces

sulfate (SO42-): ≤0.02%

cation traces

Al: ≤0.002%
Ba: ≤0.0005%
Bi: ≤0.0005%
Ca: ≤0.05%
Cd: ≤0.0005%
Co: ≤0.0005%
Cr: ≤0.0005%
Cu: ≤0.0005%
Fe: ≤0.001%
K: ≤0.010%
Li: ≤0.0005%
Mg: ≤0.002%
Mn: ≤0.0005%
Mo: ≤0.0005%
Ni: ≤0.0005%
Pb: ≤0.0005%
Sr: ≤0.002%
Zn: ≤0.0005%

absorption

≤0.08 at 280 in H2O at 0.1 M
≤0.10 at 260 in H2O at 0.1 M

SMILES string

O.[Na+].C[C@H](CCC([O-])=O)C1CCC2C3CCC4C[C@H](O)CC[C@]4(C)C3C[C@H](O)[C@]12C

InChI

1S/C24H40O4.Na.H2O/c1-14(4-9-22(27)28)18-7-8-19-17-6-5-15-12-16(25)10-11-23(15,2)20(17)13-21(26)24(18,19)3;;/h14-21,25-26H,4-13H2,1-3H3,(H,27,28);;1H2/q;+1;/p-1/t14-,15-,16-,17+,18-,19+,20+,21+,23+,24-;;/m1../s1

InChI key

NDVHNZISGVSFMP-WTCAICSISA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium deoxycholate (SDC) monohydrate is a bile acid salt.

Application

Powerful solubilization agent; used in the solubilization of estrogen synthetase from human placental microsomes.
Sodium deoxycholate monohydrate has been used in a study to assess a method for the immobilization of lipoxygenase in an alginate-silicate gel matrix. It has also been used in a study to investigate the in vitro dissolution of gall stones.

Biochem/physiol Actions

Bile Salt

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liquid crystals and phase equilibria binary bile salt-water systems.
Marques EF, et al.
Langmuir, 16(11), 5178-5186 (2000)
Immobilization of lipoxygenase in an alginate-silicate solgel matrix: formation of fatty acid hydroperoxides
Hsu, A., et al.
Biotechnology Letters, 19, 71-71 (1997)
R P Modi et al.
Journal of the Indian Medical Association, 89(5), 129-132 (1991-05-01)
An in vitro study on dissolution of 36 gall-stones obtained from 3 patients (12 stones from each patient) was carried out in heparin, bile salt and clofibrate solutions of different strengths. Heparin was found to be a poor solvent while
Effect of various lipid-bile salt mixed micelles on the intestinal absorption of amphotericin-B in rat
Dangi, J., et al.
Drug Devel. Ind. Pharm., 24, 631-631 (1998)
Catherine M Czeisler et al.
The Journal of physiology, 597(8), 2225-2251 (2019-02-02)
The embryonic PHOX2B-progenitor domain generates neuronal and glial cells which together are involved in chemosensory control of breathing and sleep homeostasis. Ablating PHOX2B-derived astrocytes significantly contributes to secondary hypoxic respiratory depression as well as abnormalities in sleep homeostasis. PHOX2B-derived astrocyte

Articles

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service