Skip to Content
Merck
All Photos(5)

Documents

C1254

Sigma-Aldrich

Sodium cholate hydrate

from bovine and/or ovine bile, ≥99%

Synonym(s):

3α,7α,12α-Trihydroxy-5β-cholan-24-oic acid sodium salt, Cholalic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H39NaO5 · xH2O
CAS Number:
Molecular Weight:
430.55 (anhydrous basis)
Beilstein:
3582354
EC Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.25

biological source

bovine and/or ovine bile

description

anionic

Assay

≥99%

form

powder

mol wt

micellar avg mol wt 900-1300

aggregation number

2-3

technique(s)

DNA purification: suitable
RNA purification: suitable
western blot: suitable

CMC

9-15 mM (20-25°C)

solubility

water: 100 mg/mL, clear, colorless to light yellow

HLB

18

SMILES string

O.[Na+].C[C@H](CCC([O-])=O)C1CCC2C3[C@H](O)CC4C[C@H](O)CC[C@]4(C)C3C[C@H](O)[C@]12C

InChI

1S/C24H40O5.Na.H2O/c1-13(4-7-21(28)29)16-5-6-17-22-18(12-20(27)24(16,17)3)23(2)9-8-15(25)10-14(23)11-19(22)26;;/h13-20,22,25-27H,4-12H2,1-3H3,(H,28,29);;1H2/q;+1;/p-1/t13-,14+,15-,16-,17+,18+,19-,20+,22+,23+,24-;;/m1../s1

InChI key

MUVVIYFKOVLQHL-RCVKHMDESA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium cholate hydrate, a non-denaturing anionic detergent, plays a crucial role in biochemical research, particularly in protein solubilization and membrane permeabilization, making it essential for protein extraction and purification, especially when dealing with membrane proteins and protein complexes. Derived from cholic acid, a prevalent human bile acid, sodium cholate hydrate′s water solubility and its applications extend to investigating the effects of various substances on cell cultures, making it a versatile tool in laboratory research.

Application

Sodium cholate hydrate has been used in a study to assess the effect of phospholipid formulation and surfactants on the dermal delivery of hydrophilic drugs from elastic liposomes. It has also been used in a study to investigate the elastic deformable vesicles, transfersomes, for transdermal drug delivery.

Biochem/physiol Actions

Bile Salt

Features and Benefits

  • Suitable for Western blotting
  • Suitable for DNA and RNA purification
  • High purity product for research applications

Other Notes

For additional information on our range of Biochemicals, please complete this form.

comparable product

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ching-Wei Lin et al.
Nature communications, 10(1), 2874-2874 (2019-06-30)
Covalent doping of single-walled carbon nanotubes (SWCNTs) can modify their optical properties, enabling applications as single-photon emitters and bio-imaging agents. We report here a simple, quick, and controllable method for preparing oxygen-doped SWCNTs with desirable emission spectra. Aqueous nanotube dispersions
Li Lei et al.
Current protocols in protein science, 84, 29-29 (2016-04-03)
Fish and human cytochrome P450 (P450) 17A1 catalyze both steroid 17α-hydroxylation and 17α,20-lyase reactions. Fish P450 17A2 catalyzes only 17α-hydroxylation. Both enzymes are microsomal-type P450s, integral membrane proteins that bind to the membrane through their N-terminal hydrophobic segment, the signal
J Abraham Domínguez-Avila et al.
Journal of the science of food and agriculture, 99(4), 1604-1612 (2018-09-07)
Serum lipoproteins are in dynamic equilibrium, partially controlled by the apolipoprotein A1 to apolipoprotein B ratio (APOA1/APOB). Freeze-dried mango pulp (FDM) is a rich source of phenolic compounds (MP) and dietary fiber (MF), although their effects on lipoprotein metabolism have
Yan Qiao et al.
Langmuir : the ACS journal of surfaces and colloids, 27(17), 11090-11097 (2011-07-19)
We report a facile method to synthesize water-soluble gold nanoparticles (AuNPs) using a biosurfactant sodium cholate as reducing reagents and protective groups in aqueous solution at ambient temperature. The diameters (13-70 nm) of uniform AuNPs can be readily adjusted by
Kevin B Ita et al.
The Journal of pharmacy and pharmacology, 59(9), 1215-1222 (2007-09-22)
The effect of phospholipid formulation and choice of surfactant on skin permeation of selected hydrophilic drugs from elastic liposomes across human epidermal membrane has been studied. Sodium cholate and various concentrations of phosphatidylcholine were used for the preparation of liposomes

Articles

Read our article about how the Nanodisc system allows for structural studies of membrane proteins.

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service