Skip to Content
Merck
All Photos(1)

Key Documents

D4875

Sigma-Aldrich

16-Dehydropregnenolone acetate

≥95%

Synonym(s):

3β-Acetoxy-5,16-pregnadien-20-one, 3β-Hydroxy-5,16-pregnadien-20-one acetate, 5,16-Pregnadien-3β-ol-20-one acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H32O3
CAS Number:
Molecular Weight:
356.50
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥95%

form

powder

SMILES string

CC(=O)OC1CCC2(C)C3CCC4(C)C(CC=C4C(C)=O)C3CC=C2C1

InChI

1S/C23H32O3/c1-14(24)19-7-8-20-18-6-5-16-13-17(26-15(2)25)9-11-22(16,3)21(18)10-12-23(19,20)4/h5,7,17-18,20-21H,6,8-13H2,1-4H3

InChI key

MZWRIOUCMXPLKV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

16-Dehydropregnenolone acetate (DPA) is synthesized from steroids sapogenin, diosgenin and solasodine. 16-Dehydropregnenolone acetate (DPA) is a crucial intermediate for the synthesis of steroid hormones-based drugs. It is an antagonist for farnesoid X receptor (FXR) and modulates cholesterol metabolism. It is considered as a potential antihyperlipidemic agent. Chemically synthesized steroid derivatives from DPA have cytotoxic features and could serve as potential anticancer agents.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A V Kamernitskiĭ et al.
Bioorganicheskaia khimiia, 33(3), 337-341 (2007-08-09)
Acetates of 3beta-hydroxy-3'-methyl-1'(N)-acylandrost-5-eno[16,17-d]pyrazolines bearing monothiooxamide acyl groups were synthesized during the study of approaches to the synthesis of 3'-methylandrosteno[16,17-d]azoles, promising biologically active analogues of 20-keto pregnenanes, and their properties were investigated. The cyclization of delta16-20-thiooxamidohydrazones to the corresponding heterocycles was
Facile green synthesis of 16-dehydropregnenolone acetate (16-DPA) from diosgenin
Baruah D, et al.
Synthetic Communications, 46(1), 79-84 (2016)
Synthesis and activity of novel 16-dehydropregnenolone acetate derivatives as inhibitors of type 1 5alpha-reductase and on cancer cell line SK-LU-1
Silva-Ortiz AV, et al.
Bioorganic & Medicinal Chemistry, 23(24), 7535-7542 (2015)
The synthesis of 16-dehydropregnenolone acetate (DPA) from potato glycoalkaloids
Vronena PJE, et al.
ARKIVOC (Gainesville, FL, United States), 2(1), 24-50 (2004)
A V Kamernitzky et al.
Journal of steroid biochemistry, 16(1), 61-67 (1982-01-01)
A new class of modified progesterones with an additional ring in the 16 alpha , 17 alpha-position (pregna-D'-pentaranes) are described. Compounds containing 4- and 6-membered D'-ring (D'4- and D'6-pentaranes) were synthesized by the cycloaddition of acetylene or 1,3-butadiene, respectively, to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service