Skip to Content
Merck
All Photos(3)

Key Documents

A8008

Sigma-Aldrich

5α-Androst-16-en-3-one

Synonym(s):

16-(5α)Androsten-3-one, 3-Keto-5α,16-androstene, Androstenone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H28O
CAS Number:
Molecular Weight:
272.43
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

Quality Level

Assay

≥98% (TLC)

form

powder

mp

140-145 °C (lit.)

solubility

dichloromethane: 9.80-10.20 mg/mL, clear, colorless to faintly yellow

shipped in

ambient

storage temp.

room temp

SMILES string

[H][C@@]12CC[C@]3([H])[C@]([H])(CC[C@]4(C)C=CC[C@@]34[H])[C@@]1(C)CCC(=O)C2

InChI

1S/C19H28O/c1-18-9-3-4-16(18)15-6-5-13-12-14(20)7-11-19(13,2)17(15)8-10-18/h3,9,13,15-17H,4-8,10-12H2,1-2H3/t13-,15-,16-,17-,18-,19-/m0/s1

InChI key

HFVMLYAGWXSTQI-QYXZOKGRSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

5α-Androst-16-en-3-one (androstenone) is a mammalian pheromone found in boar saliva, human sweat, and human urine.

Application

5α-Androst-16-en-3-one has been used:
  • to quantify its concentration in various fat tissues
  • to study its trigeminal percept and implications on the rate of specific anosmia
  • to study the effects of immunocastration on meat quality and sensory properties of pork bellies

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

G Zamaratskaia et al.
Reproduction in domestic animals = Zuchthygiene, 39(3), 168-172 (2004-06-09)
This study describes the age-related variation in boar taint compounds, skatole and androstenone, and testosterone, oestradiol-17 beta (E17 beta), oestrone sulphate (ES), dehydroepiandrosterone sulphate (DHEAS), triiodothyronine (T(3)) and insulin-like growth factor-1 (IGF-1) in six boars. Three pairs of littermates of
S Ampuero Kragten et al.
Animal : an international journal of animal bioscience, 5(10), 1634-1642 (2012-03-24)
Today, different analytical methods are used by different laboratories to quantify androstenone in fat tissue. This study shows the comparison of methods used routinely in different laboratories for androstenone quantification: Time-resolved fluoroimmunoassay in Norwegian School of Veterinary Science (NSVS; Norway)
Julie A Boyle et al.
Journal of neurobiology, 66(13), 1501-1510 (2006-10-03)
Specific anosmia is a term that describes an inability to perceive a particular odorant in the context of an otherwise normal olfactory acuity. The most common example, for the odor of androstenone, has been ascribed a prevalence ranging from 2
Maren Moe et al.
BMC genomics, 8, 405-405 (2007-11-09)
Boar taint is a major obstacle when using uncastrated male pigs for swine production. One of the main compounds causing this taint is androstenone, a pheromone produced in porcine testis. Here we use microarrays to study the expression of thousands
K M Bekaert et al.
Meat science, 88(3), 548-552 (2011-03-15)
Skatole and androstenone are the main boar taint compounds. Whereas nearly everybody is sensitive to skatole, the sensitivity to androstenone is genetically determined and differs between countries. In this study the methodology for testing androstenone sensitivity was refined and applied

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service