Skip to Content
Merck
All Photos(4)

Key Documents

D4000

Sigma-Aldrich

trans-Dehydroandrosterone

≥99%

Synonym(s):

3β-Hydroxy-5-androsten-17-one, 5-Androsten-3β-ol-17-one, DHEA, Dehydroepiandrosterone, Dehydroisoandrosterone, Prasterone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H28O2
CAS Number:
Molecular Weight:
288.42
Beilstein:
2058110
EC Number:
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77
biological source:
synthetic (organic)
form:
crystalline powder
Assay:
≥99%

biological source

synthetic (organic)

Quality Level

Assay

≥99%

form

crystalline powder

drug control

Home Office Schedule 4.2; regulated under CDSA - not available from Sigma-Aldrich Canada

mp

149-151 °C (lit.)

solubility

chloroform: 50 mg/mL, clear, colorless

shipped in

ambient

storage temp.

room temp

SMILES string

C[C@]12CC[C@H]3[C@@H](CC=C4C[C@@H](O)CC[C@]34C)[C@@H]1CCC2=O

InChI

1S/C19H28O2/c1-18-9-7-13(20)11-12(18)3-4-14-15-5-6-17(21)19(15,2)10-8-16(14)18/h3,13-16,20H,4-11H2,1-2H3/t13-,14-,15-,16-,18-,19-/m0/s1

InChI key

FMGSKLZLMKYGDP-USOAJAOKSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Dehydroandrosterone (DHEA) is a negative modulator of GABAA receptors. It is a steroid secreted by the adrenal gland with both estrogenic and androgenic properties. DHEAs conversion to 5-androstenediol, a potent estrogen, enhances the growth and survival of estrogen-sensitive breast tumors.
Negative modulator of GABAA receptors. Steroid secreted by the adrenal gland with both estrogenic and androgenic properties. Its conversion to 5-androstenediol, a potent estrogen, enhances the growth and survival of estrogen-sensitive breast tumors.
Negative modulator of GABAA receptors. Steroid secreted by the adrenal gland with both estrogenic and androgenic properties. Its conversion to 5-androstenediol, a potent estrogen, enhances the growth and survival of estrogen-sensitive breast tumors. At high but physiologically valid concentrations, DHEA inhibits proliferation of chromaffin progenitor cells and induces dopaminergic differentiation.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas Piper et al.
Drug testing and analysis, 9(11-12), 1695-1703 (2017-08-25)
In the course of investigations into the metabolism of testosterone (T) by means of deuterated T and hydrogen isotope ratio mass spectrometry, a pronounced influence of the oral administration of T on sulfoconjugated steroid metabolites was observed. Especially in case
Nikolaos E Koundourakis et al.
Hormones (Athens, Greece), 18(2), 151-172 (2019-07-01)
A constant topic reported in the lay press is the effect of sex hormones on athletic performance and their abuse by athletes in their effort to enhance their performance or to either boost or sidestep their hard, protracted, and demanding
D J Handelsman et al.
Human reproduction (Oxford, England), 32(2), 418-422 (2016-12-22)
How well does multi-analyte steroid mass spectrometry (MS) profiling classify women with and without polycystic ovary syndrome (PCOS)? Our liquid chromatography MS (LC-MS) steroid profiling only minimally improves discrimination of women with and without PCOS compared with a direct testosterone
Toshiro Migita et al.
Cancer science, 108(10), 2011-2021 (2017-08-05)
Long-chain acyl-coenzyme A (CoA) synthetase 3 (ACSL3) is an androgen-responsive gene involved in the generation of fatty acyl-CoA esters. ACSL3 is expressed in both androgen-sensitive and castration-resistant prostate cancer (CRPC). However, its role in prostate cancer remains elusive. We overexpressed
Ji Cao et al.
Molecular immunology, 112, 283-290 (2019-06-23)
Dehydroepiandrosterone (DHEA) has anti-inflammatory, anti-oxidant and immune-regulating properties, while the mechanism of DHEA actions remains unclear. The present study aims to investigate the effect and possible mechanism of DHEA on immune function of mice in vivo and in vitro. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service