D2886
DNA Ligase from T4-infected Escherichia coli
buffered aqueous glycerol solution
Synonym(s):
Polydeoxyribonucleotide Synthase, Polynucleotide Ligase, T4 DNA Ligase
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
grade
Molecular Biology
for molecular biology
form
buffered aqueous glycerol solution
specific activity
4,000 U/mL
mol wt
68 kDa
UniProt accession no.
storage temp.
−20°C
Gene Information
bacteriophage T4 ... 30(1258680)
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Suitable for:
- Ligation of blunt ended or cohesive DNA fragments
- Ligation of cloning vector and restriction insert fragments
- Seal nicks in double stranded DNA and RNA or DNA/RNA hybrids
- Couple RNA single strands by bridging oligonucleotide adapters
Biochem/physiol Actions
T4 DNA Ligase forms an energy dependent phosphodiester linkage between the termini of adjacent polynucleotides of duplex DNA. The ligation reaction requires ATP as a cofactor. Ligation of blunt-ended fragments requires higher enzyme concentration and can be facilitated by using PEG in the reaction mixture. The enzyme requires a 3′ hydroxyl and 5′ phosphate for ligation. Self-ligation of vector DNA can be prevented by dephosphorylation with alkaline phosphatase. T4 ligase plays an active role in repair of DNA and RNA nicks.
Components
T4 DNA Ligase is supplied in a solution containing 20 mM Tris-HCl (pH 7.5), 50 mM KCl, 1 mM DTT, and 50% (v/v) glycerol.
Unit Definition
One Weiss unit is defined as the amount of enzyme required to catalyze the exchange of 1 nmole of P32 from pyrophosphate into ATP as Norit-absorbable material in 20 minutes at 37°C.
Other Notes
T4 DNA Ligase is inactivated by heating at 65 °C for 10 minutes.
related product
Product No.
Description
Pricing
Glutathione S-Transferase from E. coli, recombinant, expressed in E. coli, buffered aqueous solution
Ribonuclease A from bovine pancreas, for molecular biology, ≥70 Kunitz units/mg protein, lyophilized
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Resp. Sens. 1
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Tiansheng Wang et al.
Bioorganic & medicinal chemistry letters, 22(11), 3699-3703 (2012-05-09)
A series of 4-amino-pyrido[2,3-d]pyrimidin-5(8H)-ones were designed and synthesized as a novel class of inhibitors of NAD(+)-dependent DNA ligase that possess potency against Gram-positive bacteria.
P Upcroft et al.
Gene, 51(1), 69-75 (1987-01-01)
We describe a system to generate cDNA or genomic libraries from DNA segments that have blunt termini. Background and rearrangement levels are low, but efficiencies are high and the procedural times very short. T4 ligase in the presence of polyethylene
Sambrook, J., et al.
Molecular Cloning: A Laboratory Manual, 1-1 (1989)
Ayako Nishizawa-Yokoi et al.
The New phytologist, 196(4), 1048-1059 (2012-10-12)
Evidence for the involvement of the nonhomologous end joining (NHEJ) pathway in Agrobacterium-mediated transferred DNA (T-DNA) integration into the genome of the model plant Arabidopsis remains inconclusive. Having established a rapid and highly efficient Agrobacterium-mediated transformation system in rice (Oryza
Tara Sugrue et al.
Stem cells (Dayton, Ohio), 31(1), 137-145 (2012-09-11)
The regeneration of the hematopoietic system following total body irradiation is supported by host-derived mesenchymal stromal cells (MSCs) within the bone marrow. The mechanisms used by MSCs to survive radiation doses that are lethal to the hematopoietic system are poorly
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service