Skip to Content
Merck
All Photos(1)

Key Documents

D1822

Sigma-Aldrich

Doxycycline monohydrate

Synonym(s):

6-Deoxy-5-oxytetracycline monohydrate, Doxycycline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H24N2O8 · H2O
CAS Number:
Molecular Weight:
462.45
MDL number:
UNSPSC Code:
51284030
PubChem Substance ID:
NACRES:
NA.85
Pricing and availability is not currently available.

form

powder or crystals

Quality Level

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria
mycoplasma
parasites

Mode of action

protein synthesis | interferes

storage temp.

−20°C

SMILES string

O.C[C@@H]1[C@H]2[C@H](O)[C@H]3[C@H](N(C)C)C(O)=C(C(N)=O)C(=O)[C@@]3(O)C(O)=C2C(=O)c4c(O)cccc14

InChI

1S/C22H24N2O8.H2O/c1-7-8-5-4-6-9(25)11(8)16(26)12-10(7)17(27)14-15(24(2)3)18(28)13(21(23)31)20(30)22(14,32)19(12)29;/h4-7,10,14-15,17,25,27-29,32H,1-3H3,(H2,23,31);1H2/t7-,10+,14+,15-,17-,22-;/m0./s1

InChI key

XQTWDDCIUJNLTR-CVHRZJFOSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

  • In yeast cells, doxycycline has been used in doxycycline-inducible system for overexpression of methyltransferase Hmt1 (hnRNP methyltransferase 1).[1]
  • It has been used in the doxycycline-inducible IFITM (interferon-induced transmembrane) cell lines.[2]
  • It has also been used as an antimicrobial agent.[3]

Biochem/physiol Actions

Doxycycline is a semisynthetic tetracycline that is active against bacteria, mycoplasma and protozoan parasites. It inhibits the bacterial protein synthesis by inhibiting the interaction of aminoacyl-tRNA and the bacterial ribosome.[4]

Other Notes

Keep container tightly closed in a dry and well-ventilated place.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The IFITM proteins inhibit HIV-1 infection.
Lu J, et al.
Journal of Virology, 85, 2126-2137 (2011)
Uninterpretable cerebrospinal fluid absorbance scan caused by doxycycline therapy.
Jan Miller
Annals of clinical biochemistry, 56(5), 619-622 (2019-06-07)
In vitro antimicrobial susceptibility testing of bacterial enteropathogens causing traveler's diarrhea in four geographic regions.
Gomi H, et al.
Antimicrobial Agents and Chemotherapy, 45, 212-216 (2001)
I Chopra et al.
Microbiology and molecular biology reviews : MMBR, 65(2), 232-60 (2001-05-31)
Tetracyclines were discovered in the 1940s and exhibited activity against a wide range of microorganisms including gram-positive and gram-negative bacteria, chlamydiae, mycoplasmas, rickettsiae, and protozoan parasites. They are inexpensive antibiotics, which have been used extensively in the prophlylaxis and therapy
Yeast Hmt1 catalyses asymmetric dimethylation of histone H3 arginine 2 in vitro.
Li HT, et al.
The Biochemical Journal, 467, 507-515 (2015)

Questions

1–2 of 2 Questions  
  1. What is the solubility ratio of doxycycline monohydrate D1822, and in what solvent should it be resuspended?

    1 answer
    1. The solubility is not determined for D1822 doxycycline monohydrate. An older lot 066M4022V did have a solubility specification for DMSO of 0.90 - 1.10 mg/ml. General information for the solubility of this material can be found online: Soluble in ethanol or methanol; Sparingly soluble in water; Soluble in dilute acids, alkali hydroxides, and dimethyl sulfoxide. The following article has information on the solubility of doxycycline in aqueous solution: https://www.sciencedirect.com/science/article/pii/S0022354915424694

      Helpful?

  2. Is Doxycycline monohydrate suitable for cell culture?

    1 answer
    1. This product has not been tested specifically for cell culture applications and has not been tested for endotoxin. However, a review of the peer-reviewed papers indicates that this material has been used successfully in cell culture studies. Please see the link below to review the peer-reviewed articles:
      https://www.sigmaaldrich.com/search/d1822?focus=papers&page=1&perpage=30&sort=relevance&term=D1822&type=citation_search

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service