Skip to Content
Merck
All Photos(1)

Key Documents

CS0390

Sigma-Aldrich

Mitochondria Staining Kit

1 kit sufficient for 40 tests (of 5 mL cell suspensions), 1 kit sufficient for 200 tests (of 1 mL cell suspensions)

Synonym(s):

JC-1 dye

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.32

Quality Level

usage

 kit sufficient for 200 tests (of 1 mL cell suspensions)
 kit sufficient for 40 tests (of 5 mL cell suspensions)

packaging

pkg of 1 kit

storage condition

dry at room temperature

technique(s)

flow cytometry: suitable
protein staining: suitable

fluorescence

λex 490 nm; λem 530 nm (green) (JC-1 monomers)
λex 525 nm; λem 590 nm (red) (JC-1 aggregates)

application(s)

cell analysis
detection

detection method

fluorometric

shipped in

wet ice

storage temp.

2-8°C

General description

In normal cells, the JC-1 dye concentrates in the mitochondrial matrix where it forms red fluorescent aggregates. Any event that dissipates the mitochondrial membrane potential (e.g. apoptosis) prevents the accumulation of the JC-1 dye in the mitochondria and thus, the dye is dispersed throughout the entire cell leading to a shift from red (JC-1-aggregates) to green fluorescence (JC-1 monomers). The fluorescence of the cells stained with this kit may be observed by fluorescence microscopy or measured by fluorimetric and flow cytometry analysis.

Application

The dissipation of the mitochondrial electrochemical potential gradient (Δψ) is known as an early event in apoptosis. This kit offers a fast and convenient method for the detection of changes in mitochondrial inner-membrane electrochemical potential in living cells using the cationic, lipophilic dye, JC-1.

Features and Benefits

Kit offers:
  • A fast, simple, and convenient method for staining and assaying mitochondria intactness
  • Contains all the reagents required for the detection of changes mitochondrial inner-membrane electrochemical potential
  • Contains the antibiotic valinomycin, which can be used as a control agent that prevents JC-1 aggregation
  • The shift in fluorescence is clearly detectable in both green and red channels
  • Suitable for adherent cells as well as cells in suspension
  • Has been tested with Jurkat, U-937, HeLa, NIH3T3 cells
  • The fluorescence signal can be observed by fluorescence microscopy or measured by flow cytometry

Kit Components Only

Product No.
Description

  • DMSO 1 mL

  • JC Staining Buffer 5X 120 mL

  • JC-1 1 mg

  • Valinomycin Ready Made .1 mL

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robert L Elliott et al.
PloS one, 14(4), e0214586-e0214586 (2019-04-02)
Many cells are cultured in media that contains an antibiotic to prevent bacterial contamination. Mycoplasma and other bacterial contamination is a serious problem for those involved in cell culture. Antibiotics in the media helps prevent this contamination and make life
A Cossarizza et al.
Biochemical and biophysical research communications, 197(1), 40-45 (1993-11-30)
A new method for the cytofluorimetric analysis of mitochondrial membrane potential in intact cells has been developed by using the lipophilic cationic probe 5,5',6,6'-tetrachloro-1,1',3,3'-tetraethylbenzimidazolcarbocyanine iodide (JC-1), whose monomer emits at 527 nm after excitation at 490 nm. Depending on the
Whasun Lim et al.
Biology of reproduction, 95(4), 82-82 (2016-09-02)
Luteolin is a natural compound known for its anticancer effects on various human cancers by regulating signal transduction cascades. However, the effects of luteolin on human placental choriocarcinoma are not known. Results of the present study revealed that luteolin decreased
Sunwoo Park et al.
Journal of cellular biochemistry, 119(9), 7377-7387 (2018-05-16)
Formononetin is an isoflavone that is extracted from red clovers or soy. It has anti-oxidant, anti-proliferative, and anti-tumor effects against cells in various diseases. Several cohort studies have indicated that phytoestrogen intake, including formononetin, could reduce the risk of various
Ya-Na Wu et al.
International journal of nanomedicine, 8, 3321-3331 (2013-09-17)
Previously, iron core-gold shell nanoparticles (Fe@Au) have been shown to possess cancer-preferential cytotoxicity in oral and colorectal cancer (CRC) cells. However, CRC cell lines are less sensitive to Fe@Au treatment when compared with oral cancer cell lines. In this research

Articles

Cellular oxidative stress is countered by enzymatic scavengers and antioxidant modulators against reactive oxygen species damage.

Cellular oxidative stress is countered by enzymatic scavengers and antioxidant modulators against reactive oxygen species damage.

Cellular oxidative stress is countered by enzymatic scavengers and antioxidant modulators against reactive oxygen species damage.

Cellular oxidative stress is countered by enzymatic scavengers and antioxidant modulators against reactive oxygen species damage.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service