Skip to Content
Merck
All Photos(1)

Key Documents

CLS3766

Corning® 384 well microplate, low flange

NBS (non-binding surface), black polystyrene, non-sterile, lid: no, pack of 25

Synonym(s):

384 multiwell plates, 384 well immunoassay plates, 384 well microplates, 384 well microtiter plates, 384 well plates, immunoassay plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.24
material:
black polystyrene
clear bottom
flat wells
polystyrene
maximum volume:
112 μL
size:
384 wells
working volume:
80 μL

material

black polystyrene
clear bottom
flat wells
polystyrene

sterility

non-sterile

feature

lid: no
skirt
plate format: low flange

packaging

case of 100 ea
pack of 25

manufacturer/tradename

Corning 3766

maximum volume

112 μL

size

384 wells

working volume

80 μL

color

black

binding type

NBS (non-binding surface)

Looking for similar products? Visit Product Comparison Guide

General description

Corning® 384-well ELISA Microplates with flat-bottom and low flange are high-quality, high-performance microplates that are widely used for laboratory assays. It improves assay sensitivity and performance.

Application

Corning® 384 well microplate, low flange has been used in enzyme activity assays to perform the phosphatase reactions. It has also been used to perform malachite green ATPase assays.

Features and Benefits

  • Black wall plates provide low background fluorescence, minimal light scatter, and reduced crosstalk, ensuring accurate and precise measurements
  • Thermally stable microplates (4°C to 37°C) ensure consistent performance over a wide range of temperature and storage conditions.
  • Chemically stable and noncytotoxic microplates are designed to be compatible with aqueous solutions containing low levels (<20%) of organic solvents, including ethanol and DMSO.
  • Meets SBS microplate standards to ensure compatibility with automated instruments.
  • Suitable for bar coding

Legal Information

Corning is a registered trademark of Corning, Inc.
NBS is a trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hannah Domgaard et al.
Nucleic acids research, 51(15), 8115-8132 (2023-07-03)
CRISPR-associated DinG protein (CasDinG) is essential to type IV-A CRISPR function. Here, we demonstrate that CasDinG from Pseudomonas aeruginosa strain 83 is an ATP-dependent 5'-3' DNA translocase that unwinds double-stranded (ds)DNA and RNA/DNA hybrids. The crystal structure of CasDinG reveals a
Markus Tiemann et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 28(57), e202201282-e202201282 (2022-07-06)
Discovery of protein-binding fragments for precisely defined binding sites is an unmet challenge to date. Herein, formylglycine is investigated as a molecular probe for the sensitive detection of fragments binding to a spatially defined protein site . Formylglycine peptide 3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service