Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
384 well microplate, low flange
Products
Product Category
plates (10)
Color
Manufacturer
Binding Type
Volume (µL)
Wells
Color
Plate Material
Package Size
Sterility
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
CLS3570
Corning
®
384 well microplate, low flange,
Tissue Culture (TC)-treated surface, white polystyrene, flat bottom, sterile, lid, pack of 10
Expand
View Pricing
CLS3764
Corning
®
384 well microplate, low flange,
Tissue Culture (TC)-treated surface, black polystyrene, flat bottom, sterile, lid, pack of 20
Expand
View Pricing
CLS3573
Corning
®
384 well microplate, low flange,
black polystyrene, flat bottom, non-sterile, lid: no, pack of 50
Expand
View Pricing
CLS3572
Corning
®
384 well microplate, low flange,
white polystyrene, square, non-sterile, lid: no, pack of 50
Expand
View Pricing
CLS3571
Corning
®
384 well microplate, low flange,
Tissue Culture (TC)-treated surface, black polystyrene, flat bottom, sterile, lid, pack of 10
Expand
View Pricing
CLS3577
Corning
®
384 well microplate, low flange,
high binding surface, black polystyrene, flat bottom, non-sterile, lid: no, pack of 50
Expand
View Pricing
CLS3764BC
Corning
®
384 well microplate, low flange,
Tissue Culture (TC)-treated surface, black polystyrene, flat bottom, sterile, Generic Barcode, lid, pack of 20
Expand
View Pricing
CLS3762
Corning
®
384 well microplate, low flange,
black polystyrene, flat bottom, non-sterile, lid: no, pack of 25
Expand
View Pricing
CLS3766
Corning
®
384 well microplate, low flange,
NBS
™
(non-binding surface), black polystyrene, non-sterile, lid: no, pack of 25
Expand
View Pricing
CLS3576
Corning
®
384 well microplate, low flange,
high binding surface, white polystyrene, flat bottom, non-sterile, lid: no, pack of 50
Expand
View Pricing