CL4B200
Sepharose™ CL-4B Size Exclusion Resin
Cross-linked, 90-350 mesh
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Product Name
Sepharose™ CL-4B, Cross-linked
Quality Level
form
beads
technique(s)
LPLC: suitable
matrix
Agarose
matrix active group
polymer
bead diameter
40-165 μm
pore size
30,000-5,000,000 fractionation range (Dextrans)
60,000-20,000,000 fractionation range (Globular proteins)
separation technique
size exclusion (SEC)
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
CL4B200-1L′s update product number is GE17-0150-01
Application
- Molecular size distribution assessment of Haemophilus influenzae vaccine containing lactose by HPAEC-PAD and colorimetric assays: This study used Sepharose CL-4B in a methodology assessing molecular size distribution in vaccine formulations, indicating its application in quality control and development processes in vaccine production (Nadim AH et al., 2022).
- Camel hydatidosis diagnostic kit: optimization of turnip and horseradish peroxidase conjugates using glutaraldehyde method: This research mentions the use of Sepharose CL-4B in the development of diagnostic kits, suggesting its role in enhancing diagnostic assay development through effective enzyme conjugation techniques (Kandil OM et al., 2020).
- Pectin from leaves of birch (Betula pendula Roth.): Results of NMR experiments and hypothesis of the RG-I structure: Sepharose CL-4B was utilized in the structural elucidation of pectins from birch leaves, underscoring its importance in the structural study of polysaccharides which is essential for understanding their biological functions and industrial applications (Golovchenko VV et al., 2022).
Legal Information
Sepharose is a trademark of Cytiva
related product
Product No.
Description
Pricing
replaced by
Product No.
Description
Pricing
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
100.4 - 109.4 °F
Flash Point(C)
38 - 43 °C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
S Cochet et al.
Journal of chromatography. A, 663(2), 175-186 (1994-03-11)
A range of substances were screened to find eluents for human immunoglobulin G (IgG) which are retained with a strong affinity by immobilized Drimarene Rubine R/K-5BL. The strong affinity of IgG for the dye is partly due to the presence
M M Diogo et al.
Journal of chromatography. A, 1006(1-2), 137-148 (2003-08-27)
The effect of salt and temperature on the interaction of homo-oligonucleotides with a derivatized Sepharose CL-6B hydrophobic support has been explained by the application of two different models: the solvophobic theory and the preferential interaction analysis. It has been shown
N Ubrich et al.
Artificial cells, blood substitutes, and immobilization biotechnology, 24(1), 65-75 (1996-01-01)
Immunoadsorption is an application of affinity chromatography, as a therapeutic method to specifically deplete biological fluids such as blood plasma from proteins in excess, or to extract a biomolecule from a complex mixture. However, the leakage of small amounts of
pH sensitization of phospholipid vesicles via complexation with synthetic poly(carboxylic acid)s.
D A Tirrell et al.
Annals of the New York Academy of Sciences, 446, 237-248 (1985-01-01)
M Hamai et al.
Comparative biochemistry and physiology. B, Comparative biochemistry, 95(2), 255-259 (1990-01-01)
1. Tryptic cleavage of fish myosin subfragment-1 (S-1) revealed its similar substructure of heavy chain to that of rabbit S-1. 2. The structural stability of fish S-1 was studied by thermal denaturation method, and a rapid polymerization of inactivated fish
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service