Skip to Content
Merck
All Photos(1)

Key Documents

CL2B300

Sigma-Aldrich

Sepharose CL-2B Size Exclusion Resin

Cross-linked, 70-230 mesh

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
23151817
NACRES:
NA.56

product name

Sepharose CL-2B, Cross-linked

form

suspension

bead diameter

60-200 μm

pore size

100,000-20,000,000 fractionation range (Dextrans)
70,000-40,000,000 fractionation range (Globular proteins)

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

CL2B300-1L′s updated product number is GE17-0140-01

Application

Sepharose CL-2B is used in affinity chromatography, gel filtration chromatography, protein chromatography, separation media and resins. Sepharose CL-2B has been used to study ovarian carcinoma and cystic fibrosis.
Sepharose CL-2B has been used as resins in:
  • gel permeation chromatography (GPC)
  • gravity chromatography
  • streptavidin affinity chromatography
  • size exclusion chromatography

Legal Information

Sepharose is a trademark of Cytiva

replaced by

Product No.
Description
Pricing

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

102.2 °F - closed cup

Flash Point(C)

39 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H J Grimek et al.
Biology of reproduction, 30(2), 397-409 (1984-03-01)
Acetone precipitates of bovine follicular fluid from small (3-5 mm) and large (10-20 mm) follicles were fractionated by nondissociative procedures involving chromatography on Sephacryl S-300, DEAE cellulose, and Sepharose CL-2B. Most of the proteoglycan material eluted with a relatively low
Mouse liver mitochondria isolation, size fractionation, and real-time MOMP measurement
Renault TT, et al.
Bio-protocol, 6(15) (2016)
Development of a quantitative assay for the characterization of human collectin-11 (CL-11, CL-K1)
Bayarri-Olmos R, et al.
Frontiers in immunology, 9, 2238-2238 (2018)
G. W. Cheng et al.
Plant physiology, 111(2), 447-457 (1996-06-01)
The locule tissue of tomato (Lycopersicon esculentum, Mill.) undergoes extensive liquefaction during ripening. In this study, the solubility, molecular mass, and glycosyl composition of locule pectic and alkali-soluble polysaccharides were examined with the aim of identifying features contributing to the
Evaluation of the molecular structural parameters of normal rice starch and their relationships with its thermal and digestion properties
Lin L, et al.
Molecules (Basel), 22(9), 1526-1526 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service