Skip to Content
Merck
All Photos(1)

Key Documents

COR-L23

92031919, human lung, Epithelial

Synonym(s):

COR-L23P Cells, CORL23 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

COR-L23, 92031919

biological source

human lung

growth mode

Adherent

karyotype

Very complex

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian lung large cell carcinoma

Cell Line Description

Derived from the pleural effusion of a 62-year-old Caucasian male. The cells are tumorigenic in nude mice.

Application

Studies on lung cancer

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service