Skip to Content
Merck
All Photos(1)

Key Documents

C9983

Sigma-Aldrich

Creatine Kinase BB Fraction human

≥90% (SDS-PAGE), liquid, recombinant, expressed in Pichia pastoris

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352204
eCl@ss:
32160410
NACRES:
NA.54

recombinant

expressed in Pichia pastoris

Quality Level

Assay

≥90% (SDS-PAGE)

form

liquid

mol wt

35-50 kDa

concentration

≥1.0 mg/mL

UniProt accession no.

storage temp.

−70°C

Gene Information

human ... CKB(1152)

General description

Solution in 50% glycerol, Tris buffer with 10 mM Bis-Tris-HCl, 0.5mM DTT, 0.5mM EDTA

Application

Human creatine kinase BB fraction has been used to investigate the survival benefit of the late percutaneous coronary intervention in patients after acute myocardial infarction. Human creatine kinase BB fraction has also been used in a study to analyze protein oxidations and resultant loss of function.

Biochem/physiol Actions

The product is used as a control in monitoring levels in cerebrospinal fluid by ELISA. Creatine kinase (CK) BB is a member of the CK gene family and is a cytosolic CK isoform present in the brain. The molecular mass is found to be 35-50 kDa. CKs catalyze the reversible transfer of a phosphoryl group between ATP and creatine. It plays an important role in the regulation of ATP level in neural cells. Therefore, a decrease in CK BB activity is one of the biochemical markers of the CNS cell damage in age-related neurodegenerative diseases such as Alzheimer′s disease, Pick′s disease, and Lewy body dementia.

Unit Definition

One unit will transfer 1.0 μmole of phosphate from creatine phosphate to ADP per minute at 37 °C (measured at 340 nm as one equimolar amount of NADH produced by coupled reaction).

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Talvik et al.
Acta paediatrica (Oslo, Norway : 1992), 84(10), 1183-1187 (1995-10-01)
Creatine kinase isoenzyme BB was determined in cerebrospinal fluid (CSF) in 79 preterm neonates using an original enzyme-linked immunosorbent assay. The criterion for inclusion was an Apgar score of 7 or less at 5 min of life. Neurological examination was
M Aksenov et al.
Journal of neurochemistry, 74(6), 2520-2527 (2000-05-23)
Creatine kinase (CK) BB, a member of the CK gene family, is a predominantly cytosolic CK isoform in the brain and plays a key role in regulation of the ATP level in neural cells. CK BB levels are reduced in
Bozidarka Knezević et al.
Bosnian journal of basic medical sciences, 9(1), 54-58 (2009-03-17)
The impact of late percutaneous coronary intervention (PCI) in the patients after acute myocardial infarction (AMI) on long term mortality remains to be established. At currently, thrombolysis is accepted as standard therapy when PCI is not immediately available. However, PCI
Paul D Hamilton et al.
Biochemistry and biophysics reports, 22, 100748-100748 (2020-03-11)
Creatine kinase (CK) is an energy storage enzyme that plays an important role in energy metabolism. CK/phosphocreatine functions as an energy buffer and links ATP production sites with ATP utilization sites. Several key mutations in the αA-crystallin (cryaa) and αB-crystallin
Loïc Dayon et al.
Analytical chemistry, 80(8), 2921-2931 (2008-03-04)
A new 6-plex isobaric mass tagging technology is presented, and proof of principle studies are carried out using standard protein mixtures and human cerebrospinal fluid (CSF) samples. The Tandem Mass Tags (TMT) comprise a set of structurally identical tags which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service