Skip to Content
Merck
All Photos(1)

Key Documents

27920

Sigma-Aldrich

Sodium creatine phosphate dibasic tetrahydrate

≥98.0% (NT), powder

Synonym(s):

Creatine phosphate disodium salt tetrahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2O3PNHC(=NH)N(CH3)CH2COOH · 4H2O
CAS Number:
Molecular Weight:
327.14
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

Product Name

Sodium creatine phosphate dibasic tetrahydrate, ≥98.0% (NT)

Assay

≥98.0% (NT)

form

powder

impurities

≤0.6% free creatine

solubility

H2O: 0.1 g/mL, clear, colorless

storage temp.

2-8°C

SMILES string

O.O.O.O.[Na+].[Na+].CN(CC(O)=O)C(=N)NP([O-])([O-])=O

InChI

1S/C4H10N3O5P.2Na.4H2O/c1-7(2-3(8)9)4(5)6-13(10,11)12;;;;;;/h2H2,1H3,(H,8,9)(H4,5,6,10,11,12);;;4*1H2/q;2*+1;;;;/p-2

InChI key

HUWYWJSJJDCZRQ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Sodium creatine phosphate dibasic tetrahydrate has been used:
  • for Golgi disassembly and reassembly assay
  • to regenerate ATP for fluorescence imaging
  • for the preparation of protein synthesis master mix

Substrates

Substrate for the assay of creatine kinasecitation

Other Notes

Substrate for the assay of creatine kinase

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reconstitution of the cell cycle-regulated Golgi disassembly and reassembly in a cell-free system
Tang D, et al.
Nature Protocols, 5(4), 758-758 (2010)
Single Molecule Enzymology (2011)
A reconstituted cell-free assay for the evaluation of the intrinsic activity of purified human ribosomes
Penzo M, et al.
Nature Protocols, 11(7), 1309-1309 (2016)
W. Gerhardt et al.
Methods of Enzymatic Analysis, 3, 510-510 (1983)
Sameh S M Soliman et al.
Nature microbiology, 6(3), 313-326 (2021-01-20)
Fungi of the order Mucorales cause mucormycosis, a lethal infection with an incompletely understood pathogenesis. We demonstrate that Mucorales fungi produce a toxin, which plays a central role in virulence. Polyclonal antibodies against this toxin inhibit its ability to damage

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service