Skip to Content
Merck
All Photos(1)

Key Documents

C8645

Sigma-Aldrich

Cinoxacin

Synonym(s):

1-Ethyl-1,4-dihydro-4-oxo-[1,3]dioxolo[4,5-g]cinnoline-3-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10N2O5
CAS Number:
Molecular Weight:
262.22
EC Number:
MDL number:
UNSPSC Code:
51102829
PubChem Substance ID:
NACRES:
NA.85

form

powder or crystals

antibiotic activity spectrum

Gram-negative bacteria

Mode of action

DNA synthesis | interferes
enzyme | inhibits

SMILES string

CCN1N=C(C(O)=O)C(=O)c2cc3OCOc3cc12

InChI

1S/C12H10N2O5/c1-2-14-7-4-9-8(18-5-19-9)3-6(7)11(15)10(13-14)12(16)17/h3-4H,2,5H2,1H3,(H,16,17)

InChI key

VDUWPHTZYNWKRN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: quinolone

Application

Cinoxacin was used to study the rat renal organic anion transporter 1 (OAT1). It is used to study fluoroquinolone-resistant Streptococcus pyogenes.

Biochem/physiol Actions

Cinoxacin is a synthetic antimicrobial agent that interferes with DNA replication by inhibiting DNA gyrase and topoisomerase IV (topo IV) through tight DNA binding. The mechanism of action of cinoxacin is comparable to nalidixic acid. Cinoxacin is effective against Gram-negative bacteria and is often used to treat urinary tract infections caused by E. coli, Proteus mirabilis, Proteus vulgaris and Klebsiella sp.

Other Notes

Keep container tightly closed in a dry and well-ventilated place.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M P López-Gresa et al.
Journal of inorganic biochemistry, 92(1), 65-74 (2002-09-17)
Several novel metal-quinolone compounds have been synthesized and characterized by analytical, spectroscopic and X-ray diffraction methods. The crystal structure of the four compounds, Na(2)[(Cd(Cx)3)(Cd(Cx)3(H2O))].12H2O, [Co(Cp)2(H2O)2].9H2O, [Zn(Cp)2(H2O)2].8H2O and [Cd(HCp)2(Cl)2].4H2O, is presented and discussed: HCx=1-ethyl-1,4-dihydro-4-oxo(1,3)-dioxolo(4,5-g)cinnoline-3-carboxylic acid and HCp=1-cyclopropyl-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-3-quinoline carboxylic acid. In all
A De Sarro et al.
General pharmacology, 24(6), 1393-1401 (1993-11-01)
1. The effects of some anticonvulsant drugs against seizures induced by a combined treatment with aminophylline and quinolone in genetically epilepsy-prone rat have been investigated. 2. Animals were intraperitoneally pretreated with carbamazepine, diazepam, phenobarbital, CPPene and dizocilpine or saline and
H S Brand et al.
Clinical and experimental rheumatology, 8(4), 393-395 (1990-07-01)
Explants of immature bovine articular cartilage were exposed to nalidixic acid, pipemidic acid and cinoxacin at one and ten times the human therapeutic plasma level for 7 days. Only nalidixic acid had significant effects on the chondrocyte metabolism. 20 micrograms/ml
G Mendoza-Díaz et al.
Journal of inorganic biochemistry, 64(3), 207-214 (1996-11-15)
Studies of complexation equilibria of the antibiotic anions nalidixate and cinoxacinate with [Cu(phen)]2+ and [Cu(bipy)]2+ are reported. These studies indicate that the stability of this type of complex is strongly related to the metal environment. A correlation between the stability
S Jariyawat et al.
The Journal of pharmacology and experimental therapeutics, 290(2), 672-677 (1999-07-20)
In the present study, we investigated the interactions between antibiotics, especially beta-lactam antibiotics, and rat renal organic anion transporter 1 (OAT1). [(14)C]p-Aminohippurate (PAH) uptake via OAT1 expressed in Xenopus laevis oocytes was inhibited by all of the penicillins and cephalosporins

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service