Skip to Content
Merck
All Photos(3)

Key Documents

N8878

Sigma-Aldrich

Nalidixic acid

≥98%

Synonym(s):

1,4-Dihydro-1-ethyl-7-methyl-1,8-naphthyridin-4-one-3-carboxylic acid, 1-Ethyl-1,4-dihydro-7-methyl-4-oxo-1,8-naphthyridine-3-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12N2O3
CAS Number:
Molecular Weight:
232.24
Beilstein:
750515
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

Assay

≥98%

form

powder

mp

227-229 °C (lit.)

solubility

chloroform: 50 mg/mL

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

Mode of action

DNA synthesis | interferes
enzyme | inhibits

SMILES string

CCN1C=C(C(O)=O)C(=O)c2ccc(C)nc12

InChI

1S/C12H12N2O3/c1-3-14-6-9(12(16)17)10(15)8-5-4-7(2)13-11(8)14/h4-6H,3H2,1-2H3,(H,16,17)

InChI key

MHWLWQUZZRMNGJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: quinolone

Application

Used to modulate (inhibit) bacterial DNA gyrase-dependent processes such as DNA polymerization, (ATP-dependen)t DNA supercoiling, and chromosome fragmentation.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inna Chmelnitsky et al.
Antimicrobial agents and chemotherapy, 52(8), 2962-2965 (2008-06-04)
Fourteen out of 16 carbapenem-resistant quinolone-susceptible Enterobacter cloacae isolates were found to carry qnrB2 and bla(KPC-2) genes encoded on the same plasmid. One isolate also carried the aac(6')-Ib-cr gene. Coexistence of quinolone resistance determinants and bla(KPC-2) on the same plasmid
Lucia Pallecchi et al.
Antimicrobial agents and chemotherapy, 51(8), 2720-2725 (2007-06-06)
A survey carried out in 2005 among members of a healthy population of children living in Bolivia and Peru revealed that fecal carriage of Escherichia coli strains resistant to expanded-spectrum cephalosporins was remarkably increased compared to that observed in the
Valeria Bortolaia et al.
Antimicrobial agents and chemotherapy, 54(4), 1623-1626 (2010-01-27)
We characterized 67 Escherichia coli isolates with reduced susceptibility to cefotaxime or ceftiofur obtained from healthy broilers housed in five Italian farms. The bla(CTX-M-1), bla(CTX-M-32) and bla(SHV-12) beta-lactamase genes were identified on IncI1, IncN, or IncFIB plasmids. Considerable genetic diversity
Kate S Baker et al.
The Lancet. Infectious diseases, 15(8), 913-921 (2015-05-06)
Shigellosis is an acute, severe bacterial colitis that, in high-income countries, is typically associated with travel to high-risk regions (Africa, Asia, and Latin America). Since the 1970s, shigellosis has also been reported as a sexually transmitted infection in men who
Michelle D Brazas et al.
Antimicrobial agents and chemotherapy, 51(12), 4276-4283 (2007-09-26)
With few novel antimicrobials in the pharmaceutical pipeline, resistance to the current selection of antibiotics represents a significant therapeutic challenge. Microbial persistence in subinhibitory antibiotic environments has been proposed to contribute to the development of resistance. Pseudomonas aeruginosa cultures pretreated

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service