Skip to Content
Merck
All Photos(1)

Key Documents

C5238

Sigma-Aldrich

Chlorotoxin from Leiurus quinquestriatus (north Africa)

recombinant, expressed in E. coli, ≥98% (HPLC), lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C158H256N52O48S11
CAS Number:
Molecular Weight:
4004.76
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

recombinant

expressed in E. coli

Quality Level

Assay

≥98% (HPLC)

form

lyophilized powder

solubility

aqueous buffer: soluble

storage temp.

−20°C

InChI

1S/C158H249N53O47S11/c1-77(2)55-96-138(241)205-105(144(247)186-86(125(165)228)28-18-47-173-156(166)167)70-264-262-68-103-131(234)177-63-115(217)176-64-116(218)183-87(25-12-15-44-159)128(231)178-65-117(219)184-88(29-19-48-174-157(168)169)129(232)179-66-118(220)185-89(26-13-16-45-160)133(236)202-107-72-267-269-75-110-149(252)195-98(56-81-23-10-9-11-24-81)143(246)208-124(80(5)213)154(257)209-123(79(4)212)153(256)199-102(61-122(226)227)141(244)196-99(58-83-62-172-76-181-83)139(242)189-92(37-39-113(163)215)136(239)190-94(42-53-260-7)132(235)182-78(3)126(229)187-91(30-20-49-175-158(170)171)134(237)188-90(27-14-17-46-161)135(238)203-108(73-266-265-71-106(146(249)193-96)204-137(240)93(38-40-114(164)216)191-151(254)111-31-21-50-210(111)119(221)67-180-130(233)97(194-147(107)250)57-82-33-35-84(214)36-34-82)148(251)198-100(59-120(222)223)140(243)197-101(60-121(224)225)142(245)206-109(150(253)201-103)74-268-263-69-104(200-127(230)85(162)41-52-259-6)145(248)192-95(43-54-261-8)155(258)211-51-22-32-112(211)152(255)207-110/h9-11,23-24,33-36,62,76-80,85-112,123-124,212-214H,12-22,25-32,37-61,63-75,159-162H2,1-8H3,(H2,163,215)(H2,164,216)(H2,165,228)(H,172,181)(H,176,217)(H,177,234)(H,178,231)(H,179,232)(H,180,233)(H,182,235)(H,183,218)(H,184,219)(H,185,220)(H,186,247)(H,187,229)(H,188,237)(H,189,242)(H,190,239)(H,191,254)(H,192,248)(H,193,249)(H,194,250)(H,195,252)(H,196,244)(H,197,243)(H,198,251)(H,199,256)(H,200,230)(H,201,253)(H,202,236)(H,203,238)(H,204,240)(H,205,241)(H,206,245)(H,207,255)(H,208,246)(H,209,257)(H,222,223)(H,224,225)(H,226,227)(H4,166,167,173)(H4,168,169,174)(H4,170,171,175)/t78-,79+,80+,85-,86-,87-,88-,89-,90-,91-,92-,93-,94-,95-,96-,97-,98-,99-,100-,101-,102-,103-,104-,105-,106-,107-,108-,109-,110-,111-,112-,123-,124-/m0/s1

InChI key

QPAKKWCQMHUHNI-GQIQPHNSSA-N

Amino Acid Sequence

Met-Cys-Met-Pro-Cys-Phe-Thr-Thr-Asp-His-Gln-Met-Ala-Arg-Lys-Cys-Asp-Asp-Cys-Cys-Gly-Gly-Lys-Gly-Arg-Gly-Lys-Cys-Tyr-Gly-Pro-Gln-Cys-Leu-Cys-Arg

Biochem/physiol Actions

Chlorotoxin (CLTX) is a peptide isolated from the venom of scorpion Leiurus quinquestriatus. It comprises three lysines at the N-terminal region and four disulfide bonds. It can traverse the blood-brain barrier and can associate specifically with cancer cells especially in neuroblastoma, glioma, and medulloblastoma. CLTX binds to chloride-gated channels (CLC30) and the matrix metalloproteinase-2 (MMP2) in glioma cells. It is exploited in the imaging probe preparations for aiding non-cancer and cancer cell differentiation.
Potent blocker of small-conductance Cl- channels.

Features and Benefits

This compound is featured on the Chloride Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Ullrich et al.
Neuroscience, 83(4), 1161-1173 (1998-03-21)
Using whole-cell patch-clamp recordings, we identified a novel voltage-activated chloride current that was selectively expressed in glioma cells from 23 patient biopsies. Chloride currents were identified in 64% of glioma cells studied in acute slices of nine patient biopsies. These
Joy L Kovar et al.
Analytical biochemistry, 440(2), 212-219 (2013-05-29)
Primary brain tumors present significant challenges for surgical resection because of their location and the frequent occurrence of malignant projections extending beyond the primary tumor. Visualization of the tumor margins during surgery is critical for a favorable outcome. We report
Paola G Ojeda et al.
Biopolymers, 108(5) (2017-05-02)
Chlorotoxin (CTX), a disulfide-rich peptide from the scorpion Leiurus quinquestriatus, has several promising biopharmaceutical properties, including preferential affinity for certain cancer cells, high serum stability, and cell penetration. These properties underpin its potential for use as a drug design scaffold
Dorra Aissaoui et al.
International journal of biological macromolecules, 111, 1146-1155 (2018-02-09)
Voltage-gated potassium (Kv) channels are known to play a pivotal role in the progression of various cancer types and considered as new targets for designing anti-cancer therapy. However, the fact that many Kv channels are expressed in different cell lines
Amber Emmerson et al.
The Journal of clinical investigation, 128(7), 3088-3101 (2018-04-25)
The superoxide-generating enzyme Nox2 contributes to hypertension and cardiovascular remodeling triggered by activation of the renin-angiotensin system. Multiple Nox2-expressing cells are implicated in angiotensin II-induced (Ang II-induced) pathophysiology, but the importance of Nox2 in leukocyte subsets is poorly understood. Here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service