Skip to Content
Merck
All Photos(4)

Key Documents

50950

Sigma-Aldrich

Guanidine hydrochloride

≥98%

Synonym(s):

Aminoformamidine hydrochloride, Aminomethanamidine hydrochloride, Guanidinium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2C(=NH)NH2 · HCl
CAS Number:
Molecular Weight:
95.53
Beilstein:
3591990
EC Number:
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:
NACRES:
NA.55
Pricing and availability is not currently available.

Assay

≥98%

form

powder or crystals

storage condition

(Tightly closed. Dry. )

color

colorless to white

pH

(25 °C, 4.6 - 6/573 g/L)

mp

180-185 °C (lit.)

solubility

H2O: 6 M, clear (100 Hazen)

density

1.3 g/cm3 (lit.)

SMILES string

Cl[H].NC(N)=N

InChI

1S/CH5N3.ClH/c2-1(3)4;/h(H5,2,3,4);1H

InChI key

PJJJBBJSCAKJQF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Guanidine hydrochloride, or aminoformamidine hydrochloride, is a versatile chemical compound widely used in cell biology and biochemical research due to its potent chaotropic properties. Its ability to disrupt protein and nucleic acid structures makes it valuable for purifying proteins and nucleic acids, particularly mRNA. Guanidine hydrochloride unfolds proteins, facilitating their purification and refolding, and it is effective in solubilizing denatured insoluble proteins at higher concentrations.

At lower concentrations, guanidine hydrochloride has the intriguing ability to promote the refolding of denatured proteins, aiding in protein renaturation studies. In RNA extraction, it acts as a strong denaturant, disrupting cell structures and ensuring the integrity of extracted RNA by inactivating RNA enzymes. Overall, guanidine hydrochloride′s denaturing and renaturing properties make it an essential reagent for various cell biology applications, including protein purification, nucleic acid isolation, and protein refolding studies.

Application

Guanidine hydrochloride has been used:
  • for lysing homogenized brain tissue
  • in the preparation of incubation buffer for Ni-sepharose protein binding, purification, propionylation, and on-bead digestion to minimize nonspecific binding to the affinity resin
  • in the preparation of solutions/extraction buffer to extract soluble protein from the human tissue

Strong chaotropic agent useful for the denaturation and subsequent refolding of proteins. This strong denaturant can solubilize insoluble or denatured proteins such as inclusion bodies. This can be used as the first step in refolding proteins or enzymes into their active form. Urea and dithiothreitol (DTT) may also be necessary.

Biochem/physiol Actions

Guanidine hydrochloride, an efficient chaotropic reagent, is involved in RNA isolation, globular protein denaturation, and protein refolding studies. This denaturant is appropriate for performing rapid and single-step affinity-purification mass spectrometry (AP-MS) experiments.

Features and Benefits

Highly versatile surfactant for your cell biology and biochemical research

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reyhan Yaka et al.
Current biology : CB, 31(11), 2455-2468 (2021-04-16)
The social organization of the first fully sedentary societies that emerged during the Neolithic period in Southwest Asia remains enigmatic,1 mainly because material culture studies provide limited insight into this issue. However, because Neolithic Anatolian communities often buried their dead
Andrew M King et al.
Nature communications, 12(1), 6343-6343 (2021-11-05)
Peptide secondary metabolites are common in nature and have diverse pharmacologically-relevant functions, from antibiotics to cross-kingdom signaling. Here, we present a method to design large libraries of modified peptides in Escherichia coli and screen them in vivo to identify those that bind
Satoshi Muraoka et al.
Journal of proteome research, 20(3), 1733-1743 (2021-02-04)
Extracellular vesicles (EVs) are secreted by any neural cells in the central nervous system for molecular clearance, cellular communications, and disease spread in multiple neurodegenerative diseases, including Alzheimer's disease (AD), although their exact molecular mechanism is poorly understood. We hypothesize
Christopher J Poon et al.
Bio-protocol, 7(3), e2128-e2128 (2017-02-05)
Hydrogels are an ideal medium for the expansion of cells in three dimensions. The ability to induce cell expansion and differentiation in a controlled manner is a key goal in tissue engineering. Here we describe a detailed method for producing
Armando M De Palma et al.
Antimicrobial agents and chemotherapy, 53(5), 1850-1857 (2009-02-25)
A novel compound, TTP-8307, was identified as a potent inhibitor of the replication of several rhino- and enteroviruses. TTP-8307 inhibits viral RNA synthesis in a dose-dependent manner, without affecting polyprotein synthesis and/or processing. Drug-resistant variants of coxsackievirus B3 were all

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service