Skip to Content
Merck
All Photos(3)

Key Documents

289329

Sigma-Aldrich

Cesium chloride

ReagentPlus®, 99.9%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CsCl
CAS Number:
Molecular Weight:
168.36
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.55
Assay:
99.9%
form:
crystals, chunks or beads
Pricing and availability is not currently available.

Quality Level

product line

ReagentPlus®

Assay

99.9%

form

crystals, chunks or beads

pH

6-7.5

mp

645 °C (lit.)

SMILES string

[Cl-].[Cs+]

InChI

1S/ClH.Cs/h1H;/q;+1/p-1

InChI key

AIYUHDOJVYHVIT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Cesium chloride can be used as:
  • A reagent to synthesize macrocyclic tetralactones via ring-closing metathesis reaction of diolefins in the presence of Grubbs catalyst.
  • An additive in the palladium-catalyzed selective carbonylative coupling of less reactive aryl chlorides with primary and secondary aliphatic and aromatic amines to synthesize aromatic amides.
  • An activator for low-valent titanium reagents that can be used in the reductive coupling of acetophenone.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany
Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F F White et al.
Journal of bacteriology, 164(1), 33-44 (1985-10-01)
The T-DNA regions of the root-inducing (Ri) plasmid pRiA4b of Agrobacterium rhizogenes were characterized. Two regions, designated TL-DNA and TR-DNA, were found to be integrated and stably maintained in the plant genome. The TL-DNA spanned a 15- to 20-kilobase region
Eagleson M.
Concise Encyclopedia Chemistry, 198-198 (1994)
Ribonucleic acid isolated by cesium chloride centrifugation.
Glisin V, et al.
Biochemistry, 13(12), 2633-2637 (1974)
J S Miller et al.
The Journal of clinical investigation, 84(4), 1188-1195 (1989-10-01)
The amino acid sequence of human mast cell tryptase was determined from corresponding cDNA cloned from a lambda ZAP library made with mRNA derived from a human mast cell preparation. Tryptase is the major neutral protease present in human mast
Danaé Nuzzaci et al.
Cell reports, 30(9), 3067-3078 (2020-03-05)
Mechanistic studies in rodents evidenced synaptic remodeling in neuronal circuits that control food intake. However, the physiological relevance of this process is not well defined. Here, we show that the firing activity of anorexigenic POMC neurons located in the hypothalamus

Articles

Centrifugation enables the separation of particles by sedimentation. Learn how to separate particles using a centrifuge and how to use Stokes' law to calculate the velocity of sedimentation.

Centrifugation enables the separation of particles by sedimentation. Learn how to separate particles using a centrifuge and how to use Stokes' law to calculate the velocity of sedimentation.

Centrifugation enables the separation of particles by sedimentation. Learn how to separate particles using a centrifuge and how to use Stokes' law to calculate the velocity of sedimentation.

Centrifugation enables the separation of particles by sedimentation. Learn how to separate particles using a centrifuge and how to use Stokes' law to calculate the velocity of sedimentation.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service