Skip to Content
Merck
All Photos(2)

Key Documents

C2993

Sigma-Aldrich

Anti-CRMP2 antibody produced in rabbit

enhanced validation

~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Collapsin Response Mediator Protein 2, Anti-Dihydropyriminidase Related Protein-2 (DRP2, DRP-2, DHPRP2), Anti-Dihydropyriminidase-like 2 (DPYSL2), Anti-TOAD-64

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~62 kDa

species reactivity

human, mouse, rat

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

concentration

~1.0 mg/mL

technique(s)

western blot: 0.1-0.2 μg/mL using HeLa whole cell lysate and mouse brain extract (S1 fraction)

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DPYSL2(1808)
mouse ... Dpysl2(12934)
rat ... Dpysl2(25416)

General description

Collapsin response mediator proteins CRMPs) consist of a family of cytosolic phosphoproteins expressed in the nervous system and involved in neuronal differentiation and axonal guidance.

Application

Anti-CRMP2 antibody produced in rabbit has been used in:
  • western blotting
  • immunohistofluorescence
  • epifluorescence imaging
  • coimmunoprecipitation

Biochem/physiol Actions

CRMPs are a part of the collapsin/semaphorin signal transduction pathway implicated in semaphorin-induced growth cone collapse during neural development. CRMP2 is upregulated during development, and appears to be crucial for axon outgrowth. Glycogen synthase kinase 3 beta (GSK-3b) phosphorylates and inactivates CRMP-2 downstream of the phosphoinositide-3-kinase (PI3K/Akt) pathway, thus regulating neuronal polarity. CRMP2 interacts with tubulin dimers, kinesin-1 and WASP-family verprolin homologous protein 1 (WAVE1) complex to regulate axon outgrowth.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Erik Thomas Dustrude et al.
Channels (Austin, Tex.), 11(4), 316-328 (2017-03-10)
The neuronal collapsin response mediator protein 2 (CRMP2) undergoes several posttranslational modifications that codify its functions. Most recently, CRMP2 SUMOylation (addition of small ubiquitin like modifier (SUMO)) was identified as a key regulatory step within a modification program that codes
Jennifer Y Xie et al.
Pain, 157(9), 2124-2140 (2016-08-19)
Uncoupling the protein-protein interaction between collapsin response mediator protein 2 (CRMP2) and N-type voltage-gated calcium channel (CaV2.2) with an allosteric CRMP2-derived peptide (CBD3) is antinociceptive in rodent models of inflammatory and neuropathic pain. We investigated the efficacy, duration of action
Efficacy of (S)-Lacosamide in preclinical models of cephalic pain
Moutal A, et al.
Pain reports, 1(1) (2016)
CRMP2 phosphorylation drives glioblastoma cell proliferation
Moutal A, et al.
Molecular Neurobiology, 55(5), 4403-4416 (2018)
Aubin Moutal et al.
Frontiers in cellular neuroscience, 8, 471-471 (2015-02-13)
The microtubule-associated axonal specification collapsin response mediator protein 2 (CRMP2) is a novel target for neuroprotection. A CRMP2 peptide (TAT-CBD3) conjugated to the HIV transactivator of transcription (TAT) protein's cationic cell penetrating peptide (CPP) motif protected neurons in the face

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service