Skip to Content
Merck

ABC1391

Anti-Ring Finger Protein 213 (RNF213)

from rabbit

Synonym(s):

E3 ubiquitin-protein ligase RNF213, ALK lymphoma oligomerization partner on chromosome 17, Mysterin, RING finger protein 213, RING-type E3 ubiquitin transferase RNF213

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ICC, IP, WB
Citations:
2
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse

packaging

antibody small pack of 25 μg

technique(s)

immunocytochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... RNF213(57674)

General description

E3 ubiquitin-protein ligase RNF213 (UniProt: Q63HN8; also known as ALK lymphoma oligomerization partner on chromosome 17, Mysterin, RING finger protein 213, RING-type E3 ubiquitin transferase RNF21) is encoded by the RNF213 (also known as ALO17, C17orf27, KIAA1554, KIAA1618, MYSTR) gene (Gene ID: 57674) in human. RNF213 is a widely expressed, homooligomeric, cytoplasmic protein that has E3 ubiquitin-protein ligase and AAA+ ATPase activity. It contains a zinc-finger domain (aa 3997-4036) that is required for its ubiquitin-protein ligase activity. RNF213 is shown to be involved in angiogenesis and in non-canonical Wnt signaling pathway in vascular development. Four isoforms of RNF213 have been described that are produced by alternative splicing. RNF213 ligase activity is negatively regulated by PTP1B in HER2+ breast cancer cells and RNF213 knockdown is shown to reverse the effects of PTP1B deficiency on alpha-keto-dependent dehydrogenases, non-mitochondrial oxygen consumption, and hypoxia-induced death of HER2+ BC cells. RNF213 activity is down-regulated by let-7c miRNA, which binds to the 3′-UTR transcript of RNF213. Microbial infection leading to induction of pro-inflammatory cytokines are shown to up-regulate its activity. Defects in RNF213 gene are known to cause Moyamoya disease 2, a progressive cerebral angiopathy characterized by bilateral intracranial carotid artery stenosis and telangiectatic vessels in the region of the basal ganglia, which can lead to transient ischemia and/or rupture of the collateral vessel. (Ref.: Banh, RS et al (2016). Nat. Cell Biol. 18(7); 803-813).
~596 kDa observed; 591.41 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Immunogen

11 GST-tagged recombinant fragments from human Ring Finger Protein 213.

Application

Anti-Ring Finger Protein 213 (RNF213), Cat. No. ABC1391, is a rabbit polyclonal antibody that detects E3 ubiquitin-protein ligase RNF213 and has been tested for use in Immunocytochemistry, Immunoprecipitation, and Western Blotting.
Immunoprecipitation Analysis: A representative lot detected Ring Finger Protein 213 (RNF213) in BT474, MDA-MB-361, and HCC1954 cells (Banh, R.S., et. al. (2016). Nat Cell Biol. 18(7):803-13).

Western Blotting Analysis: A 1:2,000 dilution from a representative lot detected Ring Finger Protein 213 (RNF213) in HUVEC transfected with siRNA213 (Courtesy of Dr Akio Koizumi at Kyoto University).

Immunocytochemistry Analysis: A representative lot detected Ring Finger Protein 213 (RNF213) in HeLa cells treated with RNF213 siRNA (Hitomi, T., et. al. (2013). Biochem Biophys Res Commun. 438(1):13-9).

Western Blotting Analysis: A representative lot detected Ring Finger Protein 213 (RNF213) in Western Blotting applications (Hitomi, T., et. al. (2013). Biochem Biophys Res Commun. 438(1):13-9; Banh, R.S., et. al. (2016). Nat Cell Biol. 18(7):803-13; Kobayashi, H., et. al. (2015). J Am Heart Assoc. 4(7)).

Biochem/physiol Actions

This rabbit polyclonal antibody detects human and mouse Ring Finger Protein 213.

Physical form

Format: Purified

Analysis Note

Evaluated by Western Blotting in HEK293T cells transfected with 3XFlag RNF213 wild-type.

Western Blotting Analysis: 1 µg/mL of this antibody detected Ring Finger Protein 213 (RNF213) in HEK293T cells transfected with 3XFlag RNF213 wild-type.

Other Notes

Concentration: Please refer to lot specific datasheet.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

does not flash

flash_point_c

does not flash



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library