Skip to Content
Merck
All Photos(3)

Key Documents

C2404

Sigma-Aldrich

Citric acid

anhydrous, suitable for cell culture, suitable for plant cell culture

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC(COOH)(CH2COOH)2
CAS Number:
Molecular Weight:
192.12
Beilstein:
782061
EC Number:
MDL number:
UNSPSC Code:
12161700
eCl@ss:
39021801
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

Quality Level

Assay

≥97% (GC)

form

powder

expl. lim.

8 %, 65 °F

technique(s)

cell culture | mammalian: suitable
cell culture | plant: suitable

pH

1.8 (25 °C, 50 g/L)

pKa 

(1) 3.13, (2) 4.76, (3) 6.4

mp

153-159 °C (lit.)

solubility

water: 50 mg/mL, clear to very slightly hazy, colorless

density

1.67 g/cm3 at 20 °C

SMILES string

OC(=O)CC(O)(CC(O)=O)C(O)=O

InChI

1S/C6H8O7/c7-3(8)1-6(13,5(11)12)2-4(9)10/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)

InChI key

KRKNYBCHXYNGOX-UHFFFAOYSA-N

Gene Information

human ... SRC(6714)

Looking for similar products? Visit Product Comparison Guide

Application

In addition to its function as an iron chelator, citrate can support cholesterol/sterol, ubiquinone and isoprenoid biosynthesis in cell culture applications.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria José Garcia-Fernandez et al.
Biofouling, 29(3), 261-271 (2013-02-27)
Prevention and management of wound infections receive a lot of attention, since the presence of micro-organisms interferes with the wound-healing process. The aim of this work was to use cyclodextrins (CDs) to endow hydrogels and gauzes with the ability to
Konrad Hoetzenecker et al.
European heart journal, 36(11), 676-685 (2013-01-17)
Supernatants of serum-free cultured mononuclear cells (MNC) contain a mix of immunomodulating factors (secretome), which have been shown to attenuate detrimental inflammatory responses following myocardial ischaemia. Inflammatory dilated cardiomyopathy (iDCM) is a common cause of heart failure in young patients.
Tetsuji Sawada et al.
Gastric cancer : official journal of the International Gastric Cancer Association and the Japanese Gastric Cancer Association, 18(1), 119-128 (2014-02-04)
The G-Project committee was erected by the Japan Society for Gastroenterological Carcinogenesis with an aim of establishing a new classification scheme based on molecular biological characteristics that would supplement the conventional TNM classification to better predict outcome. In a literature
Wayne B Batchelor et al.
Circulation, 106(12), 1470-1476 (2002-09-18)
The relative anti-aggregatory effects of currently prescribed platelet glycoprotein IIb/IIIa receptor antagonists during and after percutaneous coronary intervention for acute coronary syndromes have not been established. We randomized 70 acute coronary syndrome patients undergoing percutaneous coronary intervention to receive abciximab
Shihhui Huang et al.
Hippocampus, 25(8), 963-975 (2015-01-24)
Work on laboratory and wild rodents suggests that domestication may impact on the extent of adult hippocampal neurogenesis and its responsiveness to regulatory factors. There is, however, no model of laboratory rodents and their nondomesticated conspecifics that would allow a

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service