![Citric acid anhydrous, European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/162/434/37e9c009-49bc-4abc-8a8b-0f5520b5088b/640/37e9c009-49bc-4abc-8a8b-0f5520b5088b.png)
Citric acid
![Citric acid anhydrous, European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/162/434/37e9c009-49bc-4abc-8a8b-0f5520b5088b/640/37e9c009-49bc-4abc-8a8b-0f5520b5088b.png)
Synonym(s):
Citric acid
Linear Formula:
HOC(COOH)(CH2COOH)2
CAS Number:
Molecular Weight:
192.12
Products
Product Category
Brand
Sustainability
Feature
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
pK Value
Product Line
Purity
Agency
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-16 of 16
Compare
Product Number
Description
Pricing