Skip to Content
Merck
All Photos(1)

Documents

C1240

Sigma-Aldrich

Chlormethiazole hydrochloride

≥98% (HPLC), powder

Synonym(s):

4-methyl-5-(β-chloroethyl)thiazole hydrochloride, 5-(2-Chloroethyl)-4-methylthiazole hydrochloride, Clomethiazole hydrochloride, Somnevrin hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8ClNS · HCl
CAS Number:
Molecular Weight:
198.11
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

description

Store desiccated

Quality Level

Assay

≥98% (HPLC)

form

powder

color

off-white

solubility

H2O: soluble >10 mg/mL

originator

AstraZeneca

SMILES string

Cl.Cc1ncsc1CCCl

InChI

1S/C6H8ClNS.ClH/c1-5-6(2-3-7)9-4-8-5;/h4H,2-3H2,1H3;1H

InChI key

OFXYKSLKNMTBHK-UHFFFAOYSA-N

Biochem/physiol Actions

Chlormethiazole serves as an anticonvulsant. It is useful in treating alcohol addiction and eclampsia. Chlormethiazole is also useful in treating convulsive status epilepticus. Long term administration of chlormethiazole leads to addiction to the drug.
GABAA agonist; glycine receptor modulator.

Features and Benefits

This compound is a featured product for ADME Tox and Neuroscience research. Discover more featured ADME Tox and Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound was developed by AstraZeneca. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pediatric Critical Care Medicine: Basic Science And Clinical Evidence, 985-985 (2007)
A Nurse's Survival Guide to Acute Medical Emergencies (2011)
Koichi Nisijima et al.
Neurochemistry international, 43(2), 155-164 (2003-03-07)
The serotonin (5-HT) syndrome is the most serious toxic interaction of antidepressants, but no pharmacotherapy has yet been established. In the present study, we created an animal model of the 5-HT syndrome by intraperitoneally injecting rats with clorgyline (2 mg/kg)
M Gasior et al.
The Journal of pharmacology and experimental therapeutics, 295(1), 153-161 (2000-09-19)
Chlormethiazole positively modulates the gamma-aminobutyric acid (GABA)(A) receptor complex and is primarily used to treat certain life-threatening neurological events (e.g., refractory seizures and ethanol withdrawal syndrome). On account of several experimental and clinical studies reporting effectiveness against the toxic effects
Rachael M Nelson et al.
European journal of pharmacology, 452(3), 255-262 (2002-10-03)
Clomethiazole is a gamma-aminobutyric acid (GABA)-mimetic agent with anticonvulsant, sedative and neuroprotective properties. The pharmacological actions of clomethiazole that underlie its functional profile have not been fully explored, but are known to result from an interaction with the GABA(A) receptor.

Articles

Discover Bioactive Small Molecules for ADME/Tox

Discover Bioactive Small Molecules for ADME/Tox

Discover Bioactive Small Molecules for ADME/Tox

Discover Bioactive Small Molecules for ADME/Tox

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service