Skip to Content
Merck
All Photos(1)

Key Documents

C1115

Sigma-Aldrich

Chrysamine G

≥95% (HPLC), solid

Synonym(s):

4,4′-Bis(3-carboxy-4-hydroxyphenylazo)biphenyl disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H16N4Na2O6
CAS Number:
Molecular Weight:
526.41
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

solid

color

orange

solubility

DMSO: ~4 mg/mL

storage temp.

−20°C

SMILES string

[Na+].[Na+].Oc1ccc(cc1C([O-])=O)N=Nc2ccc(cc2)-c3ccc(cc3)N=Nc4ccc(O)c(c4)C([O-])=O

InChI

1S/C26H18N4O6.2Na/c31-23-11-9-19(13-21(23)25(33)34)29-27-17-5-1-15(2-6-17)16-3-7-18(8-4-16)28-30-20-10-12-24(32)22(14-20)26(35)36;;/h1-14,31-32H,(H,33,34)(H,35,36);;/q;2*+1/p-2

InChI key

AZOPGDOIOXKJRA-UHFFFAOYSA-L

General description

Chrysamine G is a carboxylic acid derivative of Congo red.

Biochem/physiol Actions

Chrysamine G can penetrate the blood-brain barrier. It is useful in quantifying cerebral amyloid β (Aβ) deposition in Alzheimer′s disease (AD) pathological studies.
New probe for ß-amyloid aggregates.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fusheng Yang et al.
The Journal of biological chemistry, 280(7), 5892-5901 (2004-12-14)
Alzheimer's disease (AD) involves amyloid beta (Abeta) accumulation, oxidative damage, and inflammation, and risk is reduced with increased antioxidant and anti-inflammatory consumption. The phenolic yellow curry pigment curcumin has potent anti-inflammatory and antioxidant activities and can suppress oxidative damage, inflammation
N A Dezutter et al.
European journal of nuclear medicine, 26(11), 1392-1399 (1999-11-07)
Chrysamine G (CG), an analogue of Congo red, is known to bind in vitro to the beta-amyloid protein (Abeta 10-43) and to homogenates of several regions of the brain of Alzheimer's disease (AD) patients. We synthesised a conjugate of 2-(acetamido)-CG
W E Klunk et al.
Neurobiology of aging, 16(4), 541-548 (1995-07-01)
Chrysamine-G (CG) is a carboxylic acid analogue of Congo red, a histologic dye which stains amyloid. CG binds to the beta-amyloid protein of Alzheimer's disease (AD) in vitro and partitions into the brain of normal mice. In this study, we
W E Klunk et al.
Neurobiology of aging, 15(6), 691-698 (1994-11-01)
This study describes the synthesis and in vitro testing of small molecule probes that may eventually prove useful as markers of amyloid deposition in living patients. The prototype agent, Chrysamine G (CG), is a derivative of Congo red. CG binds
D L Smith et al.
Neurobiology of disease, 8(6), 1017-1026 (2001-12-14)
Huntington's disease (HD) is a late onset neurodegenerative disorder caused by a CAG/polyglutamine (polyQ) repeat expansion. PolyQ aggregates can be detected in the nuclei and processes of neurons in HD patients and mouse models prior to the onset of symptoms.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service