Skip to Content
Merck
All Photos(2)

Documents

T6146

Sigma-Aldrich

Trypan Blue

powder, BioReagent, suitable for cell culture

Synonym(s):

Direct blue 14, Niagara blue reagent, blue live/dead cell assay

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C34H24N6O14S4Na4
CAS Number:
Molecular Weight:
960.81
Colour Index Number:
23850
Beilstein:
4360496
EC Number:
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.75

product line

BioReagent

Quality Level

form

powder

composition

Dye content, ~40%

packaging

glass bottle of 100 g
glass bottle of 25 g
glass bottle of 5 g

storage condition

dry at room temperature

technique(s)

cell culture | mammalian: suitable
tissue processing: suitable

mp

>300 °C (lit.)

solubility

H2O: 10 mg/mL

application(s)

cell analysis

SMILES string

[Na+].[Na+].[Na+].[Na+].Cc1cc(ccc1N=Nc2c(O)c3c(N)cc(cc3cc2S([O-])(=O)=O)S([O-])(=O)=O)-c4ccc(N=Nc5c(O)c6c(N)cc(cc6cc5S([O-])(=O)=O)S([O-])(=O)=O)c(C)c4

InChI

1S/C34H28N6O14S4.4Na/c1-15-7-17(3-5-25(15)37-39-31-27(57(49,50)51)11-19-9-21(55(43,44)45)13-23(35)29(19)33(31)41)18-4-6-26(16(2)8-18)38-40-32-28(58(52,53)54)12-20-10-22(56(46,47)48)14-24(36)30(20)34(32)42;;;;/h3-14,41-42H,35-36H2,1-2H3,(H,43,44,45)(H,46,47,48)(H,49,50,51)(H,52,53,54);;;;/q;4*+1/p-4

InChI key

GLNADSQYFUSGOU-UHFFFAOYSA-J

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

The trypan blue dye is an azo dye, derived from toluidine. It is soluble in water and has a molecular weight of 961 g/mol.

Application

Trypan blue has been used:
  • in cell viability assay
  • to determine the number of living and necrotic cells by staining with 0.2% trypan blue solution.
  • to stain plant tissue in order to to visualize cell death

Biochem/physiol Actions

The trypan blue dye is useful to differentiate the dead and live cells based on the cell′s ability to take up or exclude the dye. A viable cell is assumed to possess an intact cell membrane and hence excludes this dye and the cytoplasm appears clear. While on the other hand non-viable cells shows a blue cytoplasm.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Plant tissue trypan blue staining during phytopathogen infection.
Fernandez NB, et al.
Bio-protocol (2016)
Trypan blue exclusion test of cell viability
Strober W
Current Protocols in Immunology, 21(1), A-3B (1997)
Acanthamoeba: Biology and Pathogenesis, 85-85 (2009)
Knockdown of PSMB7 induces autophagy in cardiomyocyte cultures: possible role in endoplasmic reticulum stress
Kyrychenko VO. et al.
Pathobiology, 81(1), 8-14 (2014)
A histological analysis of lens capsules stained with trypan blue for capsulorrhexis in phacoemulsification cataract surgery
Singh AJ, et al.
Eye (London, England), 17(5), 567-567 (2003)

Articles

Cell based assays for cell proliferation (BrdU, MTT, WST1), cell viability and cytotoxicity experiments for applications in cancer, neuroscience and stem cell research.

Cell based assays for cell proliferation (BrdU, MTT, WST1), cell viability and cytotoxicity experiments for applications in cancer, neuroscience and stem cell research.

Cell based assays for cell proliferation (BrdU, MTT, WST1), cell viability and cytotoxicity experiments for applications in cancer, neuroscience and stem cell research.

Cell based assays for cell proliferation (BrdU, MTT, WST1), cell viability and cytotoxicity experiments for applications in cancer, neuroscience and stem cell research.

Protocols

3D Spheroid BME Cell Invasion Assay utilizes specialized plates and ECM for cell aggregation and spheroid formation in vitro.

3D Spheroid BME Cell Invasion Assay utilizes specialized plates and ECM for cell aggregation and spheroid formation in vitro.

3D Spheroid BME Cell Invasion Assay utilizes specialized plates and ECM for cell aggregation and spheroid formation in vitro.

3D Spheroid BME Cell Invasion Assay utilizes specialized plates and ECM for cell aggregation and spheroid formation in vitro.

Related Content

Three-dimensional (3D) printing of biological tissue is rapidly becoming an integral part of tissue engineering.

Three-dimensional (3D) printing of biological tissue is rapidly becoming an integral part of tissue engineering.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service