Skip to Content
Merck
All Photos(1)

Key Documents

B7260

Sigma-Aldrich

Nα-Benzoyl-L-arginine-7-amido-4-methylcoumarin hydrochloride

protease substrate, ≥99% (TLC), powder

Synonym(s):

Bz-Arg-AMC hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H25N5O4 · HCl
CAS Number:
Molecular Weight:
471.94
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

Product Name

Nα-Benzoyl-L-arginine-7-amido-4-methylcoumarin hydrochloride, protease substrate

Assay

≥99% (TLC)

form

powder

solubility

DMSO:methanol (1:1): 20 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

Cl.CC1=CC(=O)Oc2cc(NC(=O)[C@H](CCCNC(N)=N)NC(=O)c3ccccc3)ccc12

InChI

1S/C23H25N5O4.ClH/c1-14-12-20(29)32-19-13-16(9-10-17(14)19)27-22(31)18(8-5-11-26-23(24)25)28-21(30)15-6-3-2-4-7-15;/h2-4,6-7,9-10,12-13,18H,5,8,11H2,1H3,(H,27,31)(H,28,30)(H4,24,25,26);1H/t18-;/m0./s1

InChI key

XKAVYOJOHZLTDF-FERBBOLQSA-N

Application

Nα-Benzoyl-L-arginine-7-amido-4-methylcoumarin hydrochloride has been used:
  • as a substrate for trypsin-like proteolysis of peptide in red blood cells
  • as a substrate for midgut proteases of Pieris brassicae
  • as a substrate in screening serine protease activity of venom

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Substrates

A sensitive fluorogenic substrate for papain and trypsin.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Purification and functional characterisation of rhinocerase, a novel serine protease from the venom of Bitis gabonica rhinoceros
Vaiyapuri S, et al.
Testing, 5(3), e9687-e9687 (2010)
Trypsin-like proteolytic contamination of commercially available psa purified from human seminal fluid
Manning ML, et al.
Prostate, 72(11), 1233-1238 (2012)
Rakesh Kumar et al.
Frontiers in physiology, 6, 95-95 (2015-04-16)
Pieris brassicae L. is a serious pest of cultivated crucifers in several parts of the world. Larvae of P. brassicae also feed prolifically on garden nasturtium (Tropaeolum majus L., of the family Tropaeolaceae). Proteolytic digestion was studied in larvae feeding
Carmen Navarro-Guillén et al.
PloS one, 16(4), e0248356-e0248356 (2021-04-10)
New and more efficient methods to sustainably intensify Aquaculture production are essential to attain the seafood demand for direct human consumption in the near future. Nutrition has been identified as one strategy of early exposure that might affect animal early
Kanaoka, Y., et al.
Chemical & Pharmaceutical Bulletin, 25, 3126-3126 (1977)

Articles

Uncover properties and applications of the cysteine protease papain and find inhibitors, substrates, and other papain products.

Uncover properties and applications of the cysteine protease papain and find inhibitors, substrates, and other papain products.

Uncover properties and applications of the cysteine protease papain and find inhibitors, substrates, and other papain products.

Uncover properties and applications of the cysteine protease papain and find inhibitors, substrates, and other papain products.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service