Skip to Content
Merck
All Photos(1)

Key Documents

B6649

Sigma-Aldrich

Anti-Mouse IgG (whole molecule)−Biotin antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

biotin conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

technique(s)

direct ELISA: 1:20,000

storage temp.

2-8°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

The product binds all mouse Igs.

Application

Anti-Mouse IgG (whole molecule) antibody produced in rabbit is suitable for ELISA at a working dilution of 1:20,000. It may be used for immunohistochemistry of formalin-fixed, paraffin-embedded sections at a dilution of 1:1000.

Biochem/physiol Actions

IgG antibody subtype is the most abundant of serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defense of the neonate against infections. The coupling of biotin to Anti-Mouse IgG antibody allows for the binding of various labels such as avidin or streptavidin.

Other Notes

Antibody adsorbed with human serum proteins.

Physical form

Solution in 0.01 M phosphate buffered saline pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide.

Preparation Note

Adsorbed to reduce background with human samples.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carolina Veaute et al.
Fertility and sterility, 91(4), 1256-1268 (2008-04-29)
To assess the effect of antiacrosin antibodies upon proacrosin/acrosin activities and animal fertility. Prospective study. Basic research laboratory. A gene immunization (GI) model was developed; mice were injected with the sequence encoding human proacrosin (h-proacrosin), cloned in an expression vector.
Cecilia Pérez Brandán et al.
Memorias do Instituto Oswaldo Cruz, 114, e180571-e180571 (2019-05-23)
TcP21 is a ubiquitous secreted protein of Trypanosoma cruzi and its recombinant form (rP21) promotes parasite cell invasion and acts as a phagocytosis inducer by activating actin polymerisation in the host cell. Our goal was to evaluate if the additional
Marieta Ruseva et al.
Immunology, 127(2), 279-288 (2009-05-30)
Mannan-binding lectin (MBL) is a plasma protein implicated in innate immune defence against a broad range of microorganisms, including viruses. It is also thought that MBL plays a role in the recruitment of the specific clonal immune response. This was
Abi Karenina Funes et al.
Andrology, 11(6), 1203-1217 (2023-01-26)
Rabbits are sensitive to dietary cholesterol and rapidly develop hypercholesterolemia, leading to prominent subfertility. Sterol regulatory element-binding protein isoform 2 drives the intracellular cholesterol pathway in many tissues, including the testicles. Its abnormal regulation could be the mainly responsible for
Lucía Rodríguez-Vázquez et al.
The Journal of comparative neurology, 529(5), 1081-1096 (2020-08-14)
The current study was conducted to assess whether a single administration of 5-bromo-2'-deoxyuridine (BrdU) interferes with cell proliferation and leads to the activation of apoptotic cellular events in the prenatal cerebellum. BrdU effects across a wide range of doses (25-300 μg/g

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service