Skip to Content
Merck
All Photos(1)

Key Documents

B7264

Sigma-Aldrich

Anti-Mouse IgG (whole molecule)−Biotin antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Biotinylated anti-Mouse IgG, Goat anti-Mouse IgG

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

biotin conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

technique(s)

direct ELISA: 1:20,000

storage temp.

2-8°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Immunoglobulin G (IgG) is a glycoprotein antibody that regulates immune responses such as phagocytosis and is also involved in the development of autoimmune diseases . Mouse IgGs have four distinct isotypes, namely, IgG1, IgG2a, IgG2b, and IgG3. IgG1 regulates complement fixation in mice . Anti-Mouse IgG (whole molecule)-Biotin antibody binds all mouse immunoglobulins.

Immunogen

Purified mouse IgG

Application

Anti-Mouse IgG (whole molecule)-Biotin antibody is suitable for use in ELISA (1:5000) and immunocytochemistry (1:50).

Physical form

Solution in 0.01 M phosphate buffered saline pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matheus C Martini et al.
Archives of virology, 160(7), 1785-1790 (2015-05-09)
The ability of avian coronaviruses to replicate in mice was investigated to investigate interspecies transmission. Two inbred mouse strains (BALB/c and A/J) with different genetic backgrounds were inoculated with the avian coronavirus strains Mass and BR-I and monitored for at
Rebekah L Bower et al.
American journal of physiology. Regulatory, integrative and comparative physiology, 310(9), R788-R793 (2016-02-26)
The calcitonin receptor (CTR) is relevant to three hormonal systems: amylin, calcitonin, and calcitonin gene-related peptide (CGRP). Receptors for amylin and calcitonin are targets for treating obesity, diabetes, and bone disorders. CGRP receptors represent a target for pain and migraine.
U Greferath et al.
The Journal of comparative neurology, 301(3), 433-442 (1990-11-15)
An antibody directed against protein kinase C (PKC) was applied to various mammalian retinae. In the cat, rat, rabbit, and macaque monkey we found PKC-like immunoreactivity in bipolar cells which had the morphology of rod bipolar cells; in the rat
T Luther et al.
The American journal of pathology, 150(4), 1231-1244 (1997-04-01)
uPAR (CD87), the receptor for the urokinase-type plasminogen activator (uPA) facilitates tumor cell invasion and metastasis by focusing uPA proteolytic activity to the cell surface. As uPAR exists in various molecular forms, it is desirable to use well defined antibodies
David S Morales Herrera et al.
Heliyon, 6(7), e04331-e04331 (2020-07-17)
Nicotinamide adenine dinucleotide (NAD) is one of the central molecules involved in energy homeostasis, cellular signaling and antioxidative defense systems. Consequently, its biosynthetic pathways and transport systems are of vital importance. The nicotinamide/nicotinate mononucleotide adenylyltransferase (NMNAT), a key enzyme in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service